Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA020989

Sigma-Aldrich

Anti-ZFAT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA1485, Anti-ZFAT1, Anti-ZNF406, Anti-zinc finger and AT hook domain containing

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

EPGGDIQQEALGDQLQLVEEEFALQGVNALKEEACPGDTQLEEGRKEPEAPGEMPAPAVHLASPQAESTALPPCELETT

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFAT(57623)

General description

The gene ZFAT (zinc finger gene in AITD susceptibility region) is mapped to human chromosome 8q24. It is present in placenta, kidney, spleen, testis and peripheral blood lymphocytes.

Immunogen

zinc finger and AT hook domain containing recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZFAT (zinc finger gene in AITD susceptibility region) is a transcription factor and is suggested to be involved in lymphocyte signaling. In immune related cells, ZFAT is proposed to control apoptosis and cell survival. It also participates in endothelium organization and capillary formation. ZFAT is down-regulated in preeclampsia. Polymorphism in ZFAT is linked with severity in Hashimoto′s disease (HD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76005

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Senji Shirasawa et al.
Human molecular genetics, 13(19), 2221-2231 (2004-08-06)
Autoimmune thyroid disease (AITD) is caused by an immune response to self-thyroid antigens and has a significant genetic component. Antisense RNA transcripts have been implicated in gene regulation. Here we have identified a novel zinc-finger gene, designated ZFAT (zinc-finger gene
Sandrine Barbaux et al.
Epigenetics, 7(9), 1079-1090 (2012-08-17)
Genomic imprinting characterizes genes with a monoallelic expression, which is dependent on the parental origin of each allele. Approximately 150 imprinted genes are known to date, in humans and mice but, though computational searches have tried to extract intrinsic characteristics
Naoya Inoue et al.
Journal of clinical immunology, 32(6), 1243-1252 (2012-06-19)
Genome-wide association studies have revealed several susceptibility genes among patients with autoimmune thyroid disease (AITD), including CTLA4, PTPN22, FCRL3, and ZFAT. However, any possible association between these genes and AITD prognosis remains unknown. The objective of this study was to
Takahiro Fujimoto et al.
FEBS letters, 583(3), 568-572 (2009-01-24)
ZFAT (also known as ZNF406), originally identified as a candidate gene for autoimmune thyroid disease, encodes a zinc-finger protein, however, its function has not been elucidated. Here, we report that human ZFAT protein is expressed in peripheral B and T

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service