Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA020430

Sigma-Aldrich

Anti-CLIP2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CAP-Gly domain-containing linker protein 2, Anti-CLIP-115, Anti-CYLN2 antibody produced in rabbit, Anti-Cytoplasmic linker protein 115, Anti-Cytoplasmic linker protein 2, Anti-Williams-Beuren syndrome chromosomal region 4 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SSSISSVSSVASSVGGRPSRSGLLTETSSRYARKISGTTALQEALKEKQQHIEQLLAERDLERAEVAKATSHICEVEKEIALLKAQHEQYVAEAEEKLQRARLLVESVRKEKVDLSNQLEEERRKVEDLQFRVEEESITKGDLELTTVA

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYLN2(7461)

General description

CAP-Gly domain containing linker protein 2 (CLIP2) possesses a structural maintenance of chromosomes (Smc) domain. The gene encoding this protein is localized on human chromosome 7q11.23.

Immunogen

CAP-Gly domain-containing linker protein 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-CLIP2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CAP-Gly domain containing linker protein 2 (CLIP2) takes part in cell division, segregation of chromosomes and in attaching organelles to microtubules. CLIP2 may be useful as a marker in papillary thyroid carcinoma (PTC). Mutations in the gene encoding it have been linked with Williams-Beuren syndrome and it has been shown to be expressed in colorectal carcinomas.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78865

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Selmansberger et al.
Oncogene, 34(30), 3917-3925 (2014-10-07)
A substantial increase in papillary thyroid carcinoma (PTC) among children exposed to the radioiodine fallout has been one of the main consequences of the Chernobyl reactor accident. Recently, the investigation of PTCs from a cohort of young patients exposed to
Martin Selmansberger et al.
Carcinogenesis, 36(7), 748-756 (2015-05-10)
A previous study on papillary thyroid carcinomas (PTC) in young patients who were exposed to (131)iodine from the Chernobyl fallout revealed an exclusive gain of chromosomal band 7q11.23 in exposed cases compared to an age-matched control cohort. CLIP2, a gene
Julia Hess et al.
Proceedings of the National Academy of Sciences of the United States of America, 108(23), 9595-9600 (2011-05-25)
The main consequence of the Chernobyl accident has been an increase in papillary thyroid carcinomas (PTCs) in those exposed to radioactive fallout as young children. Our aim was to identify genomic alterations that are associated with exposure to radiation. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service