Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA019860

Sigma-Aldrich

Anti-ATAD2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CT137, Anti-DKFZp667N1320, Anti-MGC29843, Anti-MGC5254, Anti-PRO2000

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable

immunogen sequence

SQRAVTSPGQALSTVVKPLLQNTVDKILEALQRVFPHAEFRTNKTLDSDISCPLLESDLAYSDDDVPSVYENGLSQKSSHKAKDNFNFLHLNRNACYQPMSFRPRILIVGEPGFGQGSH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATAD2(29028)

General description

ATAD2 (ATPase family, AAA domain containing 2) is an evolutionarily conserved AAA+ nuclear protein consisting of a bromodomain and a double AAA+ class ATPase domain. It is localized in in the nucleus with a molecular mass of ~170kDa. Its expression has been observed in different types of cancerous cells such as human breast cancer. It is mapped to chromosome 8q24.13.

Immunogen

ATPase family AAA domain-containing protein 2 recombinant protein epitope signature tag (PrEST)

Application

Anti-ATAD2 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

ATAD2 (ATPase family, AAA domain containing 2) is associated with various cellular processes such as DNA replication, chromatin remodeling, transcription, and DNA damage. In the E2-Induced cell Proliferation and cell cycle progression, ATAD2 functions as an ERα (estrogen receptor) coactivator and facilitates E2-stimulated expression of cell cycle regulators by protein complex-remodeling phenomena. It also plays a vital role in the tumorigenesis. ATAD2 alters chromatin accessibility at MYC target loci by directly interacting with MYC as a cofactor. The chromatin accessibility further modifies transcriptional expression of the MYC gene.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74648

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gokhan Yildiz et al.
PloS one, 8(5), e64016-e64016 (2013-05-22)
Senescence is a permanent proliferation arrest in response to cell stress such as DNA damage. It contributes strongly to tissue aging and serves as a major barrier against tumor development. Most tumor cells are believed to bypass the senescence barrier
Marco Ciró et al.
Cancer research, 69(21), 8491-8498 (2009-10-22)
The E2F and MYC transcription factors are critical regulators of cell proliferation and contribute to the development of human cancers. Here, we report on the identification of a novel E2F target gene, ATAD2, the predicted protein product of which contains
June X Zou et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(46), 18067-18072 (2007-11-14)
AAA+ proteins play crucial roles in diverse biological processes via their ATPase-driven remodeling of macromolecular complexes. Here we report our identification of an evolutionarily conserved AAA+ protein, ANCCA/pro2000, endowed with a bromodomain that is strongly induced by estrogen in human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service