Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA019492

Sigma-Aldrich

Anti-RFFL antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CARP-2, Anti-Caspases-8 and -10-associated RING finger protein 2, Anti-E3 ubiquitin-protein ligase rififylin, Anti-FYVE-RING finger protein Sakura, Anti-Fring, Anti-RING finger and FYVE-like domain-containing protein 1, Anti-RING finger protein 189, Anti-RING finger protein 34-like

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

MKVKDLRDYLSLHDISTEMCREKEELVLLVLGQQPVISQEDRTRASTLSPDFPEQQAFLTQPHSSMVPPTSPNLPSSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RFFL(117584)

General description

RFFL (Ring finger and FYVE-like domain containing E3 ubiquitin protein ligase) is a caspase-associated ring protein belonging to a family of protein ubiquitin ligases. It consists of a specialized FYVE-type zinc finger domain, a cramped phosphoinositide binding pocket and a blunted membrane insertion loop.

Immunogen

E3 ubiquitin-protein ligase rififylin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RFFL (Ring finger and FYVE-like domain containing E3 ubiquitin protein ligase) is highly involved in the p53 signaling pathway. It directly interacts with the critical regulator of p53 turnover, MDM2 and it stabilizes MDM2 by inhibiting its ubiquitination. Its distinct FYVE domains also possess binding affinity to phosphatidylinositol 3-phosphate in lipid bilayers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74251

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wensheng Yang et al.
The Journal of biological chemistry, 282(5), 3273-3281 (2006-11-24)
Caspase 8/10-associated RING proteins (CARPs) are a recently described family of protein ubiquitin ligases that interact with and negatively regulate death receptor-mediated apoptosis. Because CARPs are overexpressed in cancer and their silencing reduces cell viability and sensitizes tumor cells to
Michael D Tibbetts et al.
Structure (London, England : 1993), 12(12), 2257-2263 (2004-12-04)
The caspase-associated ring proteins (CARP1 and CARP2) are distinguished from other caspase regulators by the presence of a FYVE-type zinc finger domain. FYVE-type domains are divided into two known classes: FYVE domains that specifically bind to phosphatidylinositol 3-phosphate in lipid
Wensheng Yang et al.
Cell cycle (Georgetown, Tex.), 7(5), 670-682 (2008-04-03)
CARP1 and CARP2 proteins (CARPs) are E3 ligases that target p53 as well as phospho-p53 for degradation. Because MDM2 is a critical regulator of p53 turnover, we investigated and found that CARPs associate with MDM2. We provide evidence that CARPs
Tsukasa Okiyoneda et al.
Developmental cell, 44(6), 694-708 (2018-03-06)
The peripheral protein quality control (QC) system removes non-native membrane proteins, including ΔF508-CFTR, the most common CFTR mutant in cystic fibrosis (CF), from the plasma membrane (PM) for lysosomal degradation by ubiquitination. It remains unclear how unfolded membrane proteins are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service