Skip to Content
MilliporeSigma
All Photos(11)

Key Documents

HPA019096

Sigma-Aldrich

Anti-THOC1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Nuclear matrix protein p84, Anti-THO complex subunit 1, Anti-Tho1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

VFCGRIQLFLARLFPLSEKSGLNLQSQFNLENVTVFNTNEQESTLGQKHTEDREEGMDVEEGEMGDEEAPTTCSIPIDYNLYRKFWS

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... THOC1(9984)

General description

The gene THOC1 (THO complex subunit 1) is mapped to human chromosome 18p11.32-p11.31. The protein shuttles between the cytoplasm and nucleus.

Immunogen

THO complex subunit 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-THOC1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

THOC1 (THO complex subunit 1) is a component of THO. THO is an RNP (ribonucleoprotein) complex and is also involved in formation of larger RNP complexes like TREX (transcription export). THO participates in transcriptional elongation, transcript 3′ end formation and nuclear export. THOC1 has been shown to associate with elongating RNA polymerase II, and UAP56 (spliceosome RNA helicase DDX39B). THOC1 is up-regulated in breast tumors and is correlated with metastatic state of the tumor progression. It is also linked with prostate cancer aggressiveness. On the other hand, THOC1 suppresses cell growth in lung cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73947

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Meenalakshmi Chinnam et al.
Journal of the National Cancer Institute, 106(11), doi:10-doi:10 (2014-10-10)
The majority of newly diagnosed prostate cancers will remain indolent, but distinguishing between aggressive and indolent disease is imprecise. This has led to the important clinical problem of overtreatment. THOC1 encodes a nuclear ribonucleoprotein whose expression is higher in some
Jianmei Wan et al.
Molecular medicine reports, 9(6), 2321-2327 (2014-04-01)
THO complex 1 (Thoc1) is a human nuclear matrix protein that binds to the retinoblastoma tumor suppressor retinoblastoma protein (pRb). While some studies suggest that Thoc1 has characteristics of a tumor suppressor protein, whether Thoc1 can inhibit lung cancer cell
María S Domínguez-Sánchez et al.
BMC cancer, 11, 77-77 (2011-02-19)
One key step in gene expression is the biogenesis of mRNA ribonucleoparticle complexes (mRNPs). Formation of the mRNP requires the participation of a number of conserved factors such as the THO complex. THO interacts physically and functionally with the Sub2/UAP56
Shanchun Guo et al.
Cancer research, 65(8), 3011-3016 (2005-04-19)
The biochemical pathways that are disrupted in the genesis of sporadic breast cancers remain unclear. Moreover, the present prognosticating markers used to determine the prognosis of node-negative patient leads to probabilistic results, and the eventual clinical course is far from
Fabio Gasparri et al.
FEBS letters, 574(1-3), 13-19 (2004-09-11)
P84N5 is a death domain containing protein that interacts with the tumor suppressor retinoblastoma protein and induces apoptosis. We cloned and characterized two novel alternatively spliced versions of p84N5. The p84N5 short isoform (p84N5s) lacks the death domain and does

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service