Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA019089

Sigma-Aldrich

Anti-ZC3HC1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Nuclear-interacting partner of ALK, Anti-Nuclear-interacting partner of anaplastic lymphoma kinase, Anti-Zinc finger C3HC-type protein 1, Anti-hNIPA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

CEGQAFAVGVEKNWGAVVRSPEGTPQKIRQLIDEGIAPEEGGVDAKDTSATSQSVNGSPQAEQPSLESTSKEAFFSRVETFSSLKWAGKPFELSPLVCAKYGWV

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZC3HC1(51530)

General description

The gene ZC3HC1 (zinc finger C3HC-type gene 1) is mapped to human chromosome 7q32.2. The protein localizes in the nucleus. ZC3HC1 is strongly expressed in S and G2/M phases of the cell cycle. ZC3HC1 is commonly referred as NIPA (nuclear-interacting partner of ALK).

Immunogen

Nuclear-interacting partner of ALK recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZC3HC1 (zinc finger C3HC-type protein 1) is an F-box-containing SCF (Skp1, Cullins, F-box proteins)-type E3 ligase. Cell-cycle dependent phosphorylation of ZC3HC1 controls assembly of the SCF complex. ZC3HC1 mainly participates in ubiquitination and degradation of nuclear cyclin B1, and thereby regulates mitotic entry. Polymorphism in ZC3HC1 is linked with increased carotid intima-media thickness in humans suffering from rheumatoid arthritis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74878

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel López-Mejías et al.
Arthritis research & therapy, 15(5), R152-R152 (2013-11-30)
Rheumatoid arthritis (RA) is a complex polygenic disease associated with chronic inflammation, accelerated atherosclerosis and increased cardiovascular (CV) mortality. A recent meta-analysis has described the ZC3HC1 rs11556924 polymorphism as one of the most important signals associated with coronary artery disease
Tarja Kunnas et al.
Medicine, 94(32), e1221-e1221 (2015-08-13)
In a recent genome-wide association study, the zinc finger, C3HC-type containing 1 (ZC3HC1) polymorphism was strongly associated with coronary artery disease (CAD) by an unknown mechanism. Proprotein convertase subtilisin/kexin type 9 (PCSK9) is related with CAD through low-density lipoprotein (LDL)
Florian Bassermann et al.
Cell, 122(1), 45-57 (2005-07-13)
The regulated oscillation of protein expression is an essential mechanism of cell cycle control. The SCF class of E3 ubiquitin ligases is involved in this process by targeting cell cycle regulatory proteins for degradation by the proteasome, with the F-box

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service