Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA018905

Sigma-Aldrich

Anti-ALX1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ALX homeobox protein 1, Anti-CART-1, Anti-Cartilage homeoprotein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QIQQAKSHFAATYDISVLPRTDSYPQIQNNLWAGNASGGSVVTSCMLPRDTSSCMTPYSHSPRTDSSYTGFSNHQNQFSHVPLNNFFTDSLLTGATNGHAFETKPEFERRSSSIAVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALX1(8092)

General description

The gene ALX1 (ALX homeobox protein 1) is mapped to human chromosome 12q21.3-q22. The rodent homolog of ALX1 is present in early chondrocytes and prechondrocytic mesenchymal cells. Human ALX1 is expressed in HeLa cervical carcinoma cells and cervical tissue. The protein localizes in the nucleus. ALX1 is also called as CART1 (cartilage homeoprotein 1).

Immunogen

ALX homeobox protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ALX homeobox protein 1 (ALX1) is suggested to be a transcription modulator. Hypermethylation of ALX1 is associated with shorter relapse-free survival in stage I non-small-cell lung cancer. Loss of function of ALX1 is associated with severe facial clefting and microphthalmia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84723

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Sandoval et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(32), 4140-4147 (2013-10-02)
Non-small-cell lung cancer (NSCLC) is a tumor in which only small improvements in clinical outcome have been achieved. The issue is critical for stage I patients for whom there are no available biomarkers that indicate which high-risk patients should receive
D F Gordon et al.
DNA and cell biology, 15(7), 531-541 (1996-07-01)
Homeoproteins control cell fates during development, specifying pattern formation and the ontogeny of specific tissues and organs in embryogenesis. Cart-1 cDNA was recently cloned from a rat chondrosarcoma tumor and it encodes a protein containing a paired-like homeodomain that is
Elif Uz et al.
American journal of human genetics, 86(5), 789-796 (2010-05-11)
We present an autosomal-recessive frontonasal dysplasia (FND) characterized by bilateral extreme microphthalmia, bilateral oblique facial cleft, complete cleft palate, hypertelorism, wide nasal bridge with hypoplasia of the ala nasi, and low-set, posteriorly rotated ears in two distinct families. Using Affymetrix
R L Cai
Biochemical and biophysical research communications, 250(2), 305-311 (1998-10-01)
Homeodomain proteins play important roles in animal development by controlling the expression of genes involved in determining cell fates. The recently cloned human Cart1 gene encodes a paired-class homeodomain (hCART1), whose rodent homolog is mainly expressed in early chondrocytes and
Leah M Rommereim et al.
Science signaling, 13(661) (2020-12-10)
Small, genetically determined differences in transcription [expression quantitative trait loci (eQTLs)] are implicated in complex diseases through unknown molecular mechanisms. Here, we showed that a small, persistent increase in the abundance of the innate pathogen sensor NOD1 precipitated large changes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service