Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA018828

Sigma-Aldrich

Anti-NPAP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C15ORF2, Anti-chromosome 15 open reading frame 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

TSVITSKPMNSTSVISTVTTNASAHLTSQTAVDPEVVNMDTTAPSQVVIFTSSLSSRVSSLPNSQIHCSAEQRHPGKTSVYTSPLPFIFHNTTPSFNQLFGKEATPQPKFEAPDGQPQKASLPSA

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C15orf2(23742)

General description

The gene NPAP1 (nuclear pore-associated protein 1) is mapped to human chromosome 15q. The protein localizes at the nuclear membrane. NPAP1 shows biallelic expression in the testis and monoallelic expression in fetal brain.

Immunogen

chromosome 15 open reading frame 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Nuclear pore-associated protein 1 (NPAP1) encodes a nuclear pore complex-associated protein. It is suggested to be involved in primate spermatogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74521

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karin Buiting et al.
Genomics, 89(5), 588-595 (2007-03-06)
The Prader-Willi syndrome (PWS) region contains several genes transcribed from the paternal chromosome only. We have previously identified a testis-specific gene, C15orf2, which maps between NDN and SNURF-SNRPN and is expressed from both alleles. Here we report on two novel
C Färber et al.
Genomics, 65(2), 174-183 (2000-04-28)
Prader-Willi syndrome (PWS) results from the loss of paternal contributions for a 2-Mb imprinted region on the proximal long arm of human chromosome 15. Hitherto, five paternally active genes have been identified in this region (ZNF127, NDN, MAGEL2, SNURF-SNRPN, and
Lisa C Neumann et al.
Human molecular genetics, 21(18), 4038-4048 (2012-06-15)
The Prader-Willi syndrome (PWS) region in 15q11q13 harbours a cluster of imprinted genes expressed from the paternal chromosome only. Whereas loss of function of the SNORD116 genes appears to be responsible for the major features of PWS, the role of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service