Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

HPA018668

Sigma-Aldrich

Anti-SCAF4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CTD-binding SR-like protein RA4, Anti-SFRS15, Anti-Splicing factor, arginine/serine-rich 15

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PITPPVSIPPPHTPPISIPNSTIAGINEDTTKDLSIGNPIPTVVSGARGNAESGDSVKMYGSAVPPAAPTNLPTPPVTQPVSLLGTQGVAPGPVIGLQAPSTGLLGARPGLIPLQRPPGMP

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFRS15(57466)

General description

The gene splicing factor, arginine/serine-rich 15 (SCAF4- SR-related CTD-associated factor 4) is mapped to human chromosome 21q22.1. The protein contains a CTD- (carboxyl terminal domain of RNA polymerase II) interacting domain and an RNA recognition motif.

Immunogen

Splicing factor, arginine/serine-rich 15 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Splicing factor, arginine/serine-rich 15 (SCAF4) has been proposed to regulate splicing. p63, a p53 family member, interacts with SCAF4. This interaction has been suggested to be significant for splicing of fibroblast growth factor receptor-2 isoforms. SCAF4 polymorphism is associated with diabetic nephropathy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73933

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maisaa Alwohhaib et al.
Saudi journal of kidney diseases and transplantation : an official publication of the Saudi Center for Organ Transplantation, Saudi Arabia, 25(3), 577-581 (2014-05-14)
A number of genes have been identified in diabetic nephropathy. Association between diabetes-associated nephropathy and polymorphisms in the erythropoietin (EPO) gene, variants in the superoxide dismutase 1 (SOD1) gene and plasmacytoma variant translocation 1 (PVT1) gene have been identified. The
A Yuryev et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(14), 6975-6980 (1996-07-09)
Although transcription and pre-mRNA processing are colocalized in eukaryotic nuclei, molecules linking these processes have not previously been described. We have identified four novel rat proteins by their ability to interact with the repetitive C-terminal domain (CTD) of RNA polymerase
Alexey Fomenkov et al.
The Journal of biological chemistry, 278(26), 23906-23914 (2003-04-15)
p63, a p53 family member, is required for craniofacial and limb development as well as proper skin differentiation. However, p63 mutations associated with the ankyloblepharon-ectodermal dysplasia-clefting (AEC) syndrome (Hay-Wells syndrome) were found in the p63 carboxyl-terminal region with a sterile
Roland Becker et al.
The Journal of biological chemistry, 283(33), 22659-22669 (2008-06-14)
Concomitant with RNA polymerase II (Pol II) transcription, RNA maturation factors are recruited to the carboxyl-terminal domain (CTD) of Pol II, whose phosphorylation state changes during a transcription cycle. CTD phosphorylation triggers recruitment of functionally different factors involved in RNA
Gene expression profiling and qRT-PCR expression of RRP1B, PCNT, KIF21A and ADRB2 in leucocytes of Down's syndrome subjects.
Michele Salemi et al.
Journal of genetics, 91(1), e18-e23 (2012-05-04)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service