Skip to Content
MilliporeSigma
All Photos(10)

Key Documents

HPA018535

Sigma-Aldrich

Anti-ANXA13 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Annexin A13, Anti-Annexin XIII, Anti-Annexin, intestine-specific, Anti-Annexin-13, Anti-ISA

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

RHSQSYTLSEGSQQLPKGDSQPSTVVQPLSHPSRNGEPEAPQPAKASSPQGFDVDRDAKKLNKACKGMGTN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANXA13(312)

General description

The gene annexin A13 (ANXA13) is mapped to human chromosome 8q24.12. It is a member of a family of 13 homologous phospholipid binding proteins. All annexins bind calcium, which causes a translocation from the cytosol to the cell membrane.

Immunogen

Annexin A13 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Decreased expression of annexin 13 (ANXA13) is important for apical membrane biogenesis in epithelial organization. Absence of ANXA13 perturbs epithelial cyst formation and ciliogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74475

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juha M Torkko et al.
Journal of cell science, 121(Pt 8), 1193-1203 (2008-03-20)
Epithelial cells are vital for maintaining the complex architecture and functions of organs in the body. Directed by cues from the extracellular matrix, cells polarize their surface into apical and basolateral domains, and connect by extensive cell-cell junctions to form
Juan-Manuel Iglesias et al.
Molecular biology and evolution, 19(5), 608-618 (2002-04-19)
Annexin A13 (ANXA13) is believed to be the original founder gene of the 12-member vertebrate annexin A family, and it has acquired an intestine-specific expression associated with a highly differentiated intracellular transport function. Molecular characterization of this subfamily in a
Heinz Reiske et al.
Analytical and quantitative cytology and histology, 32(2), 61-69 (2010-08-13)
To utilize a powerful new technology for target discovery, Random Homozygous Gene Perturbation (RHGP), and to identify novel targets that cause tumor cells to become chemoresistant. RHGP was used to identify and validate genetic changes that cause chemoresistance of tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service