Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA018502

Sigma-Aldrich

Anti-MTMR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Myotubularin-related protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

GRRPPRAAGGATAGSRQPSVETLDSPTGSHVEWCKQLIAATISSQISGSVTSENVSRDYKALRDGNKLAQMEEAPLFPGESIKAIVKDVMYICPFMGAVSGTLTVTDFKLYFKNVERDPH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTMR1(8776)

General description

The gene MTMR1 (myotubularin-related protein 1) is mapped to human chromosome Xq28. It belongs to protein-tyrosine phosphatase family. The protein is localized in the cytoplasm with prominent presence also in large membrane projections and at the cell periphery.

Immunogen

Myotubularin-related protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Myotubularin-related protein-1 (MTMR1) uses phosphatidylinositol 3-phosphate (PI3P) as a substrate. The protein can modify the levels of endosomal PI3P. Mutation in MTMR1 is associated with genetic disorder, X-linked myotubular myopathy. Dysregulation of MTMR1 is associated with muscle pathological features in juvenile/adult onset myotonic dystrophy type-1 and in myotonic dystrophy type-2.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74274

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soo-A Kim et al.
The Journal of biological chemistry, 277(6), 4526-4531 (2001-12-26)
Myotubularin is the archetype of a family of highly conserved protein-tyrosine phosphatase-like enzymes. The myotubularin gene, MTM1, is mutated in the genetic disorder, X-linked myotubular myopathy. We and others have previously shown that myotubularin utilizes the lipid second messenger, phosphatidylinositol
Massimo Santoro et al.
Experimental and molecular pathology, 89(2), 158-168 (2010-08-06)
Among genes abnormally expressed in myotonic dystrophy type1 (DM1), the myotubularin-related 1 gene (MTMR1) was related to impaired muscle differentiation. Therefore, we analyzed MTMR1 expression in correlation with CUG-binding protein1 (CUG-BP1) and muscleblind-like1 protein (MBNL1) steady-state levels and with morphological
P Kioschis et al.
Genomics, 54(2), 256-266 (1998-11-26)
MTM1 is responsible for X-linked recessive myotubular myopathy, which is a congenital muscle disorder linked to Xq28. MTM1 is highly conserved from yeast to humans. A number of related genes also exist. The MTM1 gene family contains a consensus sequence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service