Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA018439

Sigma-Aldrich

Anti-IPO8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Imp8, Anti-Importin-8, Anti-Ran-binding protein 8, Anti-RanBP8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LLLHTLERIQLPHNPGPITVQFINQWMNDTDCFLGHHDRKMCIIGLSILLELQNRPPAVDAVVGQIVPSILFLFLGLKQVCATRQLVNREDRSKAEKADMEENE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IPO8(10526)

General description

The gene importin-8 (IPO8) is mapped to human chromosome 12p11.21. It is a RanGTP-binding protein and associates with nuclear protein complex at the nuclear envelope.

Immunogen

Importin-8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Importin-8 (IPO8) is mainly involved in nuclear import of proteins. Knockdown of IPO8 reduces nuclear import of p65 following tumor necrosis factor-α stimulation, resulting in lower NF-κB activity. IPO8 is required for transforming growth factor -β-induced nuclear import of SMAD4 (mothers against decapentaplegic homolog-4). The signal recognition particle (SRP) assembles in the nucleus and is required for targeting of proteins to the rough endoplasmic reticulum. IPO8 is involved in nuclear import of SRP19, a subunit of mammalian SRP. IPO8 interacts and co-localizes with argonaute (AGO) proteins to cytoplasmic processing bodies in a Ran-dependent manner. IPO8 is important for binding of AGO proteins to mRNA targets. In addition, IPO8 controls nuclear localization of AGO2.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73963

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peizhou Liang et al.
Traffic (Copenhagen, Denmark), 14(11), 1132-1143 (2013-08-03)
NF-κB/p65 is retained in the cytoplasm until it is activated in response to stress. Nuclear import of p65 is regulated by importin α in a nuclear localization signal (NLS)-dependent manner. However, the role of importin β family members in the
D Görlich et al.
The Journal of cell biology, 138(1), 65-80 (1997-07-14)
The importin-alpha/beta complex and the GTPase Ran mediate nuclear import of proteins with a classical nuclear localization signal. Although Ran has been implicated also in a variety of other processes, such as cell cycle progression, a direct function of Ran
Xiaohao Yao et al.
The Journal of biological chemistry, 283(33), 22867-22874 (2008-06-04)
Trafficking of Smad proteins between the cytoplasm and nucleus is a critical component of transforming growth factor beta (TGF-beta) signal transduction. Smad4 translocates into the nucleus either in response to TGF-beta stimulation or when its nuclear export is blocked by
K A Dean et al.
Journal of cell science, 114(Pt 19), 3479-3485 (2001-10-30)
The signal recognition particle (SRP) is a cytoplasmic RNA-protein complex that targets proteins to the rough endoplasmic reticulum. Although SRP functions in the cytoplasm, RNA microinjection and cDNA transfection experiments in animal cells, as well as genetic analyses in yeast
Paul A Nguewa et al.
BMC molecular biology, 9, 103-103 (2008-11-19)
The accurate normalization of differentially expressed genes in lung cancer is essential for the identification of novel therapeutic targets and biomarkers by real time RT-PCR and microarrays. Although classical "housekeeping" genes, such as GAPDH, HPRT1, and beta-actin have been widely

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service