Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA017979

Sigma-Aldrich

Anti-SLC30A2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Solute carrier family 30 member 2, Anti-Zinc transporter 2, Anti-ZnT-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20-1:50

immunogen sequence

VLMEGTPKGVDFTAVRDLLLSVEGVEALHSLHIWALTVAQPVLSVHIAIAQNTDAQAVLKTASSRLQGKFHFHTVTIQIEDYSEDMKDCQACQGPSD

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... SLC30A2(7780)

General description

Solute carrier family 30 member 2 (SLC30A2) is a 372 amino acid protein expressed in mammary and prostate glands. It exists as two isoforms-the 42kDa isoform which is located in the endosomal compartment and the 35kDa isoform which is present in the plasma membrane.

Immunogen

Zinc transporter 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 30 member 2 (SLC30A2) plays an important role in the maintenance of cellular zinc levels and its homeostasis. It is involved in the transportation of zinc into organelles and secretory vesicles. SLC30A2 also aids in the accumulation of zinc in mitochondria and in turn leads to increased mitochondrial oxidation. A mutation in the maternal gene encoding SLC30A2 has been linked with transient neonatal zinc deficiency (TNZD) in infants.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73139.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeynep Bostanci et al.
Experimental cell research, 321(2), 190-200 (2013-12-18)
Prolactin receptor (PRL-R) activation regulates cell differentiation, proliferation, cell survival and motility of breast cells. Prolactin (PRL) and PRL-R over-expression are strongly implicated in breast cancer, particularly contributing to tumor growth and invasion in the more aggressive estrogen-receptor negative (ER-)
Miguel Lova Navarro et al.
Pediatric dermatology, 31(2), 251-252 (2014-01-25)
Transient neonatal zinc deficiency (TNZD) has a clinical presentation similar to that of acrodermatitis enteropathica but is caused by a low zinc concentration in maternal breast milk. TNZD becomes clinically evident during breastfeeding and is resolved by weaning and the
Veronica Lopez et al.
The Biochemical journal, 422(1), 43-52 (2009-06-06)
ZnT2 (zinc transporter-2) expression is restricted to tissues with unique zinc requirements such as mammary and prostate glands. We previously determined that ZnT2 plays a major role in zinc export from mammary glands, as women with a mutation in the
Inbal Lasry et al.
The Journal of biological chemistry, 287(35), 29348-29361 (2012-06-27)
Zinc is an essential mineral, and infants are particularly vulnerable to zinc deficiency as they require large amounts of zinc for their normal growth and development. We have recently described the first loss-of-function mutation (H54R) in the zinc transporter ZnT-2
Monika Schweigel-Röntgen
Current topics in membranes, 73, 321-355 (2014-04-22)
The solute carriers families 30 (SLC30; ZnT), 39 (SLC39; ZIP), and 31 (SLC31; CTR) are involved in the essential maintenance of cellular zinc (Zn²⁺) and copper (Cu²⁺) homeostasis, respectively. ZnTs mediate Zn²⁺ extrusion from cells (SLC30A1) or transport Zn²⁺ into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service