Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA017929

Sigma-Aldrich

Anti-VRK1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Serine/threonine-protein kinase VRK1, Anti-Vaccinia-related kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

RNAi knockdown
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

LPWEDNLKDPKYVRDSKIRYRENIASLMDKCFPEKNKPGEIAKYMETVKLLDYTEKPLYENLRDILLQGLKAIGSKDDGKLDLSVVENGGLKAKTITKKRKKEIEESKEPGVEDTEWSNTQTEEAIQTRSRTR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VRK1(7443)

General description

Vaccinia-related kinase 1 (VRK1) is a nucleosomal kinase belonging to a group of serine/threonine kinases which are associated with breast carcinomas. The gene encoding VRK1 is located on human chromosome 14.

Immunogen

Serine/threonine-protein kinase VRK1 recombinant protein epitope signature tag (PrEST)

Application

Anti-VRK1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting.[1] To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Vaccinia-related kinase 1 (VRK1) phosphorylates many transcription factors like tumor suppressor p53 and c-Jun (a kinase). It controls cell-cycle progression wherein it is required for exit from G0 phase and entry into G1 phase. VRK1 interacts with proteins like histones of the chromatin network and is essential for chromatin condensation. It has been shown to phosphorylate histone H3. VRK1 plays an important role in response to DNA damage by UV-light and radiations.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70405

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcella Salzano et al.
Oncotarget, 5(7), 1770-1778 (2014-04-16)
Vaccinia-related kinase 1 (VRK1) belongs to a group of sixteen kinases associated to a poorer prognosis in human breast carcinomas, particularly in estrogen receptor positive cases based on gene expression arrays. In this work we have studied the potential molecular
Claudia Gonzaga-Jauregui et al.
JAMA neurology, 70(12), 1491-1498 (2013-10-16)
Patients with rare diseases and complex clinical presentations represent a challenge for clinical diagnostics. Genomic approaches are allowing the identification of novel variants in genes for very rare disorders, enabling a molecular diagnosis. Genomics is also revealing a phenotypic expansion
Lara El-Bazzal et al.
Human molecular genetics, 28(14), 2378-2394 (2019-05-16)
Distal hereditary motor neuropathies (dHMNs) are a heterogeneous group of diseases, resembling Charcot-Marie-Tooth syndromes, but characterized by an exclusive involvement of the motor part of the peripheral nervous system. Here, we describe two new compound heterozygous mutations in VRK1, the
Diana M Monsalve et al.
Biochimica et biophysica acta, 1863(4), 760-769 (2016-02-13)
NBS1 is an early component in DNA-Damage Response (DDR) that participates in the initiation of the responses aiming to repair double-strand breaks caused by different mechanisms. Early steps in DDR have to react to local alterations in chromatin that are
Joon Shin et al.
The Journal of biological chemistry, 286(25), 22131-22138 (2011-05-06)
Vaccinia-related kinase 1 (VRK1) is one of the mitotic kinases that play important roles in cell cycle, nuclear condensation, and transcription regulation. Kinase domain structures of two other VRK family members (VRK2 and VRK3) have been determined previously. However, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service