Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA017910

Sigma-Aldrich

Anti-RFFL antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CARP-2, Anti-Caspase regulator CARP2, Anti-Caspases-8 and -10-associated RING finger protein 2, Anti-E3 ubiquitin-protein ligase rififylin, Anti-FYVE-RING finger protein Sakura, Anti-Fring, Anti-RING finger and FYVE-like domain-containing protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL

immunogen sequence

AQATSVPPAQVQENQQANGHVSQDQEEPVYLESVARVPAEDETQSIDSEDSFVPGRRASLSDLTDLEDIEGLTVRQLKEILARNF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RFFL(117584)

General description

Ring finger and FYVE-like domain containing E3 ubiquitin protein ligase (RFFL) contains a zinc finger domain, a pocket for phosphoinositide binding and a loop which helps in membrane insertion. RFFL is localized to endocytic vesicles.

Immunogen

E3 ubiquitin-protein ligase rififylin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Ring finger and FYVE-like domain containing E3 ubiquitin protein ligase (RFFL) poly-ubiquitinates and destabilizes proline-rich 5-like (PRR5L). It also targets tumor suppressor p53 for degradation. RFFL functions as a negative regulator of tumor necrosis factor (TNF)-induced NF-κB (NF-κβ) activation by interacting with the internalized TNF-receptor complex.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74252

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wentao Liao et al.
Current biology : CB, 18(9), 641-649 (2008-05-03)
The proinflammatory cytokine tumor necrosis factor-alpha (TNF-alpha) elicits cellular responses by signaling through a receptor complex that includes the essential adaptor molecule RIP. One important consequence of signaling is activation of the transcription factor NF-kappaB, and failure to downregulate TNF-induced
Wensheng Yang et al.
Cell cycle (Georgetown, Tex.), 7(5), 670-682 (2008-04-03)
CARP1 and CARP2 proteins (CARPs) are E3 ligases that target p53 as well as phospho-p53 for degradation. Because MDM2 is a critical regulator of p53 turnover, we investigated and found that CARPs associate with MDM2. We provide evidence that CARPs
Michael D Tibbetts et al.
Structure (London, England : 1993), 12(12), 2257-2263 (2004-12-04)
The caspase-associated ring proteins (CARP1 and CARP2) are distinguished from other caspase regulators by the presence of a FYVE-type zinc finger domain. FYVE-type domains are divided into two known classes: FYVE domains that specifically bind to phosphatidylinositol 3-phosphate in lipid
Xiaoqing Gan et al.
The Journal of biological chemistry, 288(47), 33978-33984 (2013-10-12)
We previously characterized a Gα12-specific signaling pathway that stimulates the transcription of the E3 ligase RFFL via the protein kinase ARAF and ERK. This pathway leads to persistent PKC activation and is important for sustaining fibroblast migration. However, questions remain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service