Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA017265

Sigma-Aldrich

Anti-CYP4F11 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CYPIVF11, Anti-Cytochrome P450 4F11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

YDNCRRLQCFPQPPKQNWFWGHQGLVTPTEEGMKTLTQLVTTYPQGFKLWLGPTFPLLILCHPDIIRPITSASAAVAPKDMIFYGFLKPWLGDGLLLSGGDKWSRHRRMLTPAF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP4F11(57834)

General description

CYP4F11 (cytochrome P450, family 4, subfamily F, polypeptide 11) is a cytochrome P450 4F isoform belonging to the CYP4 family and CYP4A subfamily. In humans, it is expressed in liver, heart, kidney, and skeletal muscles.

Immunogen

Cytochrome P450 4F11 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CYP4F11 (cytochrome P450, family 4, subfamily F, polypeptide 11) stimulates the ω-hydroxylation reaction of 3-hydroxystearate by acting as a predominant catalyst. It has been reported that CYP4F11-promoted hydroxylation may influence fatty acid mobilization. CYP4F11 oxidizes several drugs such as erythromycin, benzphetamine, ethylmorphine, chlorpromazine and imipramine.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72250

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Auinash Kalsotra et al.
Toxicology and applied pharmacology, 199(3), 295-304 (2004-09-15)
We previously reported the cDNA cloning of a new CYP4F isoform, CYP4F11. In the present study, we have expressed CYP4F11 in Saccharomyces cerevisiae and examined its catalytic properties towards endogenous eicosanoids as well as some clinically relevant drugs. CYP4F3A, also
Madhurima Dhar et al.
Journal of lipid research, 49(3), 612-624 (2007-12-11)
Long-chain 3-hydroxydicarboxylic acids (3-OHDCAs) are thought to arise via beta-oxidation of the corresponding dicarboxylic acids (DCAs), although long-chain DCAs are neither readily transported into nor beta-oxidized in mitochondria. We thus examined whether omega-hydroxylation of 3-hydroxy fatty acids (3-OHFAs), formed via
X Cui et al.
Genomics, 68(2), 161-166 (2000-08-31)
By a combination of cDNA library screening and rapid amplification of cDNA ends analysis, a novel human cytochrome P450 4F isoform has been cloned and sequenced. The new 4F isoform is designated CYP4F11 and contains 1765 nucleotides. The coding region
A E Simpson
General pharmacology, 28(3), 351-359 (1997-03-01)
1. The CYP4 family consists of 11 subfamilies (CYP4A-CYP4M), which encode constitutive and inducible isozymes expressed in both mammals and insects. 2. The CYP4A subfamily encodes several cytochrome P450 enzymes that are capable of hydroxylating the terminal omega-carbon and, to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service