Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA016697

Sigma-Aldrich

Anti-RYR2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

RYR2 Antibody - Anti-RYR2 antibody produced in rabbit, Ryr2 Antibody, Anti-Cardiac muscle ryanodine receptor-calcium release channel, Anti-Cardiac muscle-type ryanodine receptor, Anti-RYR-2, Anti-RyR2, Anti-Ryanodine receptor 2, Anti-hRYR-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

FSKTVAGGLPGAGLFGPKNDLEDYDADSDFEVLMKTAHGHLVPDRVDKDKEATKPEFNNHKDYAQEKPSRLKQRFLLRRTKPDYSTSHSARLTEDVLADDRDDYDFLMQTSTYYYSVR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... RYR2(6262)

General description

RYR2 (ryanodine receptor 2) is the calcium release channel present on the sarcoplasmic reticulum (SR). It forms tetramer with four types 2 RyR polypeptides (RyR2) and four FK506 binding proteins (FKBP12.6).[1]

Immunogen

Ryanodine receptor 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RYR2 (ryanodine receptor 2) mediates the release and transport of Ca2+ from sarcoplasmic reticulum (SR) to the cytoplasm during cardiac muscle excitation-contraction (EC) coupling. Protein kinase A (PKA) phosphorylated RYR2 separates FKBP12.6 to regulate the channel open probability (Po).[1] Missense mutations in this gene cause Kazakh idiopathic ventricular tachycardia and arrhythmogenic right ventricular dysplasia.[2][3]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73318.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Wang et al.
American journal of physiology. Heart and circulatory physiology, 307(9), H1317-H1326 (2014-09-07)
Expression silencing of junctophilin-2 (JPH2) in mouse heart leads to ryanodine receptor type 2 (RyR2)-mediated sarcoplasmic reticulum (SR) Ca(2+) leak and rapid development of heart failure. The mechanism and physiological significance of JPH2 in regulating RyR2-mediated SR Ca(2+) leak remains
Limian Zheng et al.
Biochimica et biophysica acta. Molecular cell research, 1869(1), 119139-119139 (2021-10-09)
Trophoblasts are specialized epithelial cells of the placenta that are involved in invasion, communication and the exchange of materials between the mother and fetus. Cytoplasmic Ca2+ ([Ca2+]c) plays critical roles in regulating such processes in other cell types, but relatively
Christopher M Lewarchik et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(5), G574-G581 (2014-07-12)
Physiological calcium (Ca(2+)) signals within the pancreatic acinar cell regulate enzyme secretion, whereas aberrant Ca(2+) signals are associated with acinar cell injury. We have previously identified the ryanodine receptor (RyR), a Ca(2+) release channel on the endoplasmic reticulum, as a
Ainur Akilzhanova et al.
PloS one, 9(6), e101059-e101059 (2014-07-01)
Channelopathies, caused by disturbed potassium or calcium ion management in cardiac myocytes are a major cause of heart failure and sudden cardiac death worldwide. The human ryanodine receptor 2 (RYR2) is one of the key players tightly regulating calcium efflux
Nathalie Roux-Buisson et al.
Heart rhythm, 11(11), 1999-2009 (2014-07-22)
Arrhythmogenic right ventricular cardiomyopathy/dysplasia (ARVC/D) is a genetic disease predominantly caused by desmosomal gene mutations that account for only ~50% of cases. Ryanodine receptor 2 (RYR2) gene mutations usually cause catecholaminergic polymorphic ventricular tachycardia but have been associated with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service