Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA015113

Sigma-Aldrich

Anti-CD109 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-150 KDa TGF-beta-1-binding protein, Anti-C3 and PZP-like alpha-2- macroglobulin domain-containing protein 7, Anti-CD109 antigen precursor, Anti-Platelet-specific Gov antigen, Anti-p180, Anti-r150

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

GPVEILTTVTESVTGISRNVSTNVFFKQHDYIIEFFDYTTVLKPSLNFTATVKVTRADGNQLTLEERRNNVVITVTQRNYTEYWSGSNSGNQKMEAVQKINYTVPQSGTFKIEFPILEDSSELQLKAYFLGSKSSMA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD109(135228)

General description

CD109 is a member of a family of thioester-containing proteins, called α2-macroglobulin/complement family. It was initially recognized as a leukemia antigen, and is a glycophosphatidylinositol (GPI)-anchored glycoprotein. It is expressed by endothelial cells, platelets, activated T-cells, keratinocytes and certain CD34+ hematopoietic stem and progenitor cells. It forms a part of the TGF-β1 receptor complex.[1] This protein has a molecular weight of 180kDa.[2]

Immunogen

CD109 antigen precursor recombinant protein epitope signature tag (PrEST)

application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The function of CD109 is not well understood.[2] It suppresses the TGF-β/Smad signaling, by promoting the degradation of TGFβ receptor. In scleroderma, it prevents the production of extracellular matrix. It is expressed in esophagus, lung, breast, cervix and urinary tract cancers, and facilitates tumorigenesis in oral cancer. It acts as an antigen for acute myeloid leukemia, and is expressed at the cell surface. It is up-regulated in multiple sarcomas, and has potential as a prognostic marker for epithelioid sarcoma.[1] It is up-regulated in triple-negative breast cancer, and has potential as a marker for the progression and differentiation of breast cancer cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71508

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji Tao et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(12), 12083-12090 (2014-08-26)
The aim of this study is to explore the expression of CD109 in breast cancer stem cells and the relationship between CD109 protein and clinicopathological characteristics of breast cancer. CD44+/CD24- tumor cells (CSCs) were selected by flow cytometry. The protein
Sang Mee Hwang et al.
BioMed research international, 2013, 946403-946403 (2013-03-20)
CD109 gene encodes a glycosylphosphatidylinositol-linked glycoprotein found in a subset of platelets and endothelial cell, and human platelet antigen (HPA) 15 is found on CD109. We evaluated the HPA genotype and/or the CD109 mRNA expression on two peripheral blood stem
Makoto Emori et al.
PloS one, 8(12), e84187-e84187 (2014-01-01)
Epithelioid sarcoma (ES) is a relatively rare, highly malignant soft tissue sarcoma. The mainstay of treatment is resection or amputation. Currently other therapeutic options available for this disease are limited. Therefore, a novel therapeutic option needs to be developed. In
Xiao-Yong Man et al.
Arthritis research & therapy, 14(3), R144-R144 (2012-06-15)
Scleroderma or systemic sclerosis (SSc) is a complex connective tissue disease characterized by fibrosis of skin and internal organs. Transforming growth factor beta (TGF-β) plays a key role in the pathogenesis of SSc fibrosis. We have previously identified CD109 as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service