Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA014865

Sigma-Aldrich

Anti-ZBTB38 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CIBZ, Anti-FLJ35036, Anti-PPP1R171, Anti-ZNF921

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: 0.25-2 μg/mL

immunogen sequence

PVLSLSNSSENAASVISYSGSAPSVIVHSSQFSSVIMHSNAIAAMTSSNHRAFSDPAVSQSLKDDSKPEPDKVGRFASRPKSIKEKKKTTSHTRGEIPEESNYVADPGGSLSKTTNIAEETSKIET

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZBTB38(253461)

General description

ZBTB38 (zinc finger and BTB domain containing 38) is a transcriptional repressor, which is methyl-dependent. This gene is localized to human chromosome 3q23, contains eight exons and is ~126kb in size. It is conserved across vertebrates. The encoded protein is composed of 1196 amino acids, and is composed of zinc finger domains, a Ctbr-binding domain and a BTB-binding domain. It is highly expressed in brain, and is expressed in a tissue-specific manner.

Immunogen

Zinc finger and BTB domain-containing protein 38 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZBTB38 (zinc finger and BTB domain containing 38) represses the transcription of multiple genes, by binding to methylated DNA regions. It can also associate with sequences containing a single methylated CpG. It is cleaved by caspase 3, and its levels fall during early apoptosis. Thus, it plays a role in controlling apoptosis. Studies show that this gene is linked to human height, by mediating the repression of apoptosis-related genes. Studies in Chinese Han population show that variants in this gene are associated with idiopathic short stature. RBBP6, an E3 ubiquitin DNA ligase, ubiquitinates and causes the destabilization of ZBTB38. These proteins, along with MCM10, control the replication of DNA and chromosomal stability.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73338

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benoit Miotto et al.
Cell reports, 7(2), 575-587 (2014-04-15)
Faithful DNA replication is essential for the maintenance of genome integrity. Incomplete genome replication leads to DNA breaks and chromosomal rearrangements, which are causal factors in cancer and other human diseases. Despite their importance, the molecular mechanisms that control human
Ying Wang et al.
Clinical endocrinology, 79(3), 402-408 (2013-01-11)
Idiopathic short stature (ISS) refers to extreme short stature without any diagnostic explanation. Recently, three genome-wide association studies discovered associations between the ZBTB38 and adult height in different populations. Therefore, variations in the ZBTB38 might contribute to ISS. Furthermore, one
Kyung-Won Hong et al.
Biochemical genetics, 49(5-6), 283-291 (2010-12-29)
Recently, genome-wide association studies have identified a strong association between the ZBTB38 locus and human height. In a functional study, we detected two RT-PCR products of ZBTB38, amplified with primers in exons 7 and 8 from a chondrocyte cell line
Guillaume J P Filion et al.
Molecular and cellular biology, 26(1), 169-181 (2005-12-16)
In vertebrates, densely methylated DNA is associated with inactive transcription. Actors in this process include proteins of the MBD family that can recognize methylated CpGs and repress transcription. Kaiso, a structurally unrelated protein, has also been shown to bind methylated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service