Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA014845

Sigma-Aldrich

Anti-MPDU1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Mannose-P-dolichol utilization defect 1 protein, Anti-SL15, Anti-Suppressor of Lec15 and Lec35 glycosylation mutation homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human, mouse

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

AEADGPLKRLLVPILLPEKCYDQLFVQWDLLHVPCLKILLSK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

glycosylation

Gene Information

human ... MPDU1(9526)

General description

MPDU1 (mannose-P-dolichol utilization defect 1) is a human homolog of Lec35, which is a hamster protein. This gene is localized to human chromosome 17p12-13. The encoded transmembrane protein, spans the membrane twice, and has a molecular weight of 27kDa. It has a putative ER (endoplasmic reticulum)-retention sequence (KKXX) in its cytosolic C-terminal. The N-terminal is also thought to face the cytosol.

Immunogen

Mannose-P-dolichol utilization defect 1 protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MPDU1 (mannose-P-dolichol utilization defect 1) is responsible for the bioavailability of dolichol-P-glucose and dolichol-P-mannose (MPD), in the RER (rough endoplasmic reticulum). MPD is essential for the synthesis of glycosylphosphatidylinositols and lipid-linked oligosaccharides. These, in turn, regulate membrane anchoring and protein folding, respectively. Mutation in this gene is responsible for congenital disorders of glycosylation (CDGs), which is an inherited disorder caused due to errors in protein and lipid glycosylation. It might also be a sorting carrier of flippases, and might be responsible for either bringing or retaining flippases in the ER.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73280

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vladimir Saudek
PloS one, 7(2), e30876-e30876 (2012-03-01)
Classification of proteins into families based on remote homology often helps prediction of their biological function. Here we describe prediction of protein cargo receptors involved in vesicle formation and protein trafficking. Hidden Markov model profile-to-profile searches in protein databases using
C Kranz et al.
The Journal of clinical investigation, 108(11), 1613-1619 (2001-12-26)
We describe a new congenital disorder of glycosylation, CDG-If. The patient has severe psychomotor retardation, seizures, failure to thrive, dry skin and scaling with erythroderma, and impaired vision. CDG-If is caused by a defect in the gene MPDU1, the human
M Anand et al.
Molecular biology of the cell, 12(2), 487-501 (2001-02-17)
The Lec35 gene product (Lec35p) is required for utilization of the mannose donor mannose-P-dolichol (MPD) in synthesis of both lipid-linked oligosaccharides (LLOs) and glycosylphosphatidylinositols, which are important for functions such as protein folding and membrane anchoring, respectively. The hamster Lec35

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service