Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA014768

Sigma-Aldrich

Anti-PRLH antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PrRP, Anti-Prolactin-releasing hormone, Anti-Prolactin-releasing peptide precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

RTHRHSMEIRTPDINPAWYASRGIRPVGRFGRRRATLGDVPKPGLRPRLTCFPLEGGAMSSQD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRLH(51052)

General description

PRLH (prolactin releasing hormone) is a peptide hormone, which was initially isolated from bovine hypothalamus. It is transcribed as a preprotein, which is then cleaved into two different peptides PrRP (Prolactin-releasing peptide)31 and PrRP20. It is predominantly expressed in the hypothalamus. PRLH mRNA is also found in placenta and decidua. It has the RF-motif present at its C-terminal. It is the ligand for GPR10.

Immunogen

Prolactin-releasing peptide precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-PRLH antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PRLH (prolactin releasing hormone) acts through a G-protein coupled receptor, and induces the production of prolactin from the pituitary gland. It is expressed throughout pregnancy, by placenta and decidua and is thought have some physiological functions during pregnancy. It might be responsible for modulating the release of prolactin from the pituitary gland of the fetus. It is up-regulated in pheochromocytomas, and might be involved in tumorigenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71638

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Lagerström et al.
Annals of the New York Academy of Sciences, 1040, 368-370 (2005-05-14)
The prolactin-releasing hormone (PRLH) is implicated in food intake and is expressed in several parts of the mammalian brain. The origin of the peptide precursor (PRH) has been unclear, and the only feature resembling other known human neuropeptide sequences is
Kazuhiro Takahashi et al.
Peptides, 23(6), 1135-1140 (2002-07-20)
Adrenal tumors, such as pheochromocytomas, are known to express various peptides and their receptors. Prolactin-releasing peptide (PrRP) is a novel neuropeptide isolated from bovine hypothalamic tissues. In the present study, expression of PrRP receptor was studied in the human brain
R Fujii et al.
Regulatory peptides, 83(1), 1-10 (1999-09-25)
Prolactin-releasing peptide (PrRP) is a novel bioactive peptide, originally isolated from bovine hypothalamus by utilizing an orphan seven-transmembrane-domain receptor expressed in the human pituitary gland. In this paper, we analyzed the tissue distribution of rat and human PrRP and their
Y Yasui et al.
Endocrine journal, 48(3), 397-401 (2001-08-29)
The aims of this study were to determine whether the human placenta and decidua express PRL-releasing peptide (PrRP) mRNA and whether PrRP regulates PRL secretion from cultured human decidual cells. PrRP gene expression was analyzed by reverse transcription (RT)-PCR, and
X Zhang et al.
The Journal of clinical endocrinology and metabolism, 84(12), 4652-4655 (1999-12-22)
The recently identified PRL-releasing peptide (PrRP) is the first hypothalamic peptide hormone that specifically stimulates PRL production from the pituitary gland. Similar to other hypothalamic regulatory hormones, it acts through its specific seven-transmembrane domain, G protein-coupled receptor. Using RT-PCR, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service