Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA014743

Sigma-Aldrich

Anti-ARL6IP6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ADP-ribosylation factor-like protein 6-interacting protein 6, Anti-ARL-6- interacting protein 6, Anti-Aip-6, Anti-Phosphonoformate immuno-associated protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MSFAESGWRSALRRRGPGTPGPVARPSYSSFTQGDSWGEGEVDEEEGCDQVARDLRAEFSAGAWSEPRKRSVLPPDGNGSPVLPDKRNGIFP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

ARL6IP6 (ADP-ribosylation factor-like 6 interacting protein 6) is one of the six ARL-6 interacting proteins. It shows high sequence identity to a putative cytoskeletal protein called JWA/ ARL6IP5.

Immunogen

ADP-ribosylation factor-like 6 interacting protein 6

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The function of ARL6IP6 (ADP-ribosylation factor-like 6 interacting protein 6) is not yet characterized. A homozygous mutation leading to truncated ARL6IP6 is associated with syndromic form of Cutis Marmorata Telangiectatica Congenita (CMTC), which is characterized by severe dysmorphism, delay in development, transient ischemic attacks and vascular malformations in cerebrum.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73078

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iman S Abumansour et al.
Human genetics, 134(8), 815-822 (2015-05-11)
Cutis Marmorata Telangiectatica Congenita (CMTC) is a congenital localized or generalized vascular anomaly, usually sporadic in occurrence. It can be associated with other cutaneous or systemic manifestations. About 300 cases have been reported. The molecular etiology remains largely unknown. The
Alexander Talishinsky et al.
PloS one, 7(8), e44236-e44236 (2012-09-07)
The lateral septum has strong efferent projections to hypothalamic and midbrain regions, and has been associated with modulation of social behavior, anxiety, fear conditioning, memory-related behaviors, and the mesolimbic reward pathways. Understanding natural variation of lateral septal anatomy and function
E Ingley et al.
FEBS letters, 459(1), 69-74 (1999-10-06)
We report here the isolation of a new member of the ADP-ribosylation factor (ARF)-like family (ARL-6) present in the J2E erythroleukemic cell line, but not its myeloid variants. Consistent with this lineage-restricted expression, ARL-6 mRNA increased with erythropoietin-induced maturation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service