Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA014468

Sigma-Aldrich

Anti-APCDD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Adenomatosis polyposis coli down-regulated 1 protein, Anti-Protein APCDD1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

immunogen sequence

FHDNNNTWEGHYYHYSDPVCKHPTFSIYARGRYSRGVLSSRVMGGTEFVFKVNHMKVTPMDAATASLLNVFNGNECGAEGSWQVGIQQDVTHTNGCVALGIKLPHTEYEIFKMEQ

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APCDD1(147495)

General description

The gene APCDD1 (adenomatosis polyposis coli down-regulated 1) encodes a glycoprotein that is membrane-bound and is expressed mainly in human hair follicles. It is also abundantly expressed in adult heart, pancreas, prostate, and ovary. The gene is mapped to human chromosome 18p11.22. The gene spans a length of 40kb and contains five exons. The encoded protein is 514-amino acid long and has a molecular mass of 58.8kDa.

Immunogen

Protein APCDD1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene APCDD1 (adenomatosis polyposis coli down-regulated 1) encodes a protein that functions as an inhibitor of the Wnt signaling pathway. It is targeted by β-catenin/Tcf4 (transcription factor 4) complex and is involved in cell proliferation. The expression of this protein is found to be increased in colon cancers and may serve as a diagnostic marker. Polymorphism in this gene is associated with hereditary hypotrichosis simplex, characterized by normal hair at birth and a gradual hair loss beginning in early childhood.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72074

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Synonymous Polymorphism of APCDD1 Affects Translation Efficacy and is Associated with Androgenic Alopecia.
Nagy N
Journal of Life Sciences, 8 (2014)
Yutaka Shimomura et al.
Nature, 464(7291), 1043-1047 (2010-04-16)
Hereditary hypotrichosis simplex is a rare autosomal dominant form of hair loss characterized by hair follicle miniaturization. Using genetic linkage analysis, we mapped a new locus for the disease to chromosome 18p11.22, and identified a mutation (Leu9Arg) in the adenomatosis
Meiko Takahashi et al.
Cancer research, 62(20), 5651-5656 (2002-10-18)
To clarify the molecular mechanisms of human carcinogenesis associated with abnormal beta-catenin/T-cell factor (Tcf) signaling, we have been using cDNA microarrays to search for genes whose expression is significantly altered after introduction of wild-type APC into SW480 colon cancer cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service