Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA014261

Sigma-Aldrich

Anti-GABRA2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GABA(A) receptor subunit alpha-2, Anti-Gamma-aminobutyric acid receptor subunit alpha-2 precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

immunogen sequence

AWDGKSVVNDKKKEKASVMIQNNAYAVAVANYAPNLSKDPVLSTISKSATTPEPNKKPENKPAEAKKTFNSVSKI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GABRA2(2555)

General description

The gene GABRA2 (γ-aminobutyric acid type A receptor α2 subunit) encodes the α subunit of the GABAA receptor. It is one of the units of the cluster of four GABAA receptor genes, GABRG1, GABRA2, GABRA4, and GABRB1. These receptors are mediators of effects of alcohol and modulators of brain oscillations in the β frequencies. GABRA2 may be involved in modulation of level of neural excitation and may play a role in susceptibility to alcohol dependence. The gene cluster is localized to human chromosome 4p. The ligand, GABA, binds to the GABAA receptor and activates an integral chloride-ion channel, which leads to a change in the membrane potential of neurons.

Immunogen

Gamma-aminobutyric acid receptor subunit alpha-2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73134.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Howard J Edenberg et al.
American journal of human genetics, 74(4), 705-714 (2004-03-17)
Alcoholism is a complex disease with both genetic and environmental risk factors. To identify genes that affect the risk for alcoholism, we systematically ascertained and carefully assessed individuals in families with multiple alcoholics. Linkage and association analyses suggested that a
Sunantha Kosonsiriluk et al.
Neuroendocrinology, 103(6), 678-692 (2015-11-13)
Photoperiod is a major environmental cue in temperate-zone birds which synchronizes breeding with the time of year that offers the optimal environment for offspring survival. Despite continued long photoperiods, these birds eventually become refractory to the stimulating photoperiod and their
Josh Neman et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(3), 984-989 (2014-01-08)
Dispersion of tumors throughout the body is a neoplastic process responsible for the vast majority of deaths from cancer. Despite disseminating to distant organs as malignant scouts, most tumor cells fail to remain viable after their arrival. The physiologic microenvironment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service