Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA014133

Sigma-Aldrich

Anti-PREB antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Mammalian guanine nucleotide exchange factor mSec12, Anti-Prolactin regulatory element-binding protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

LRFQAHQQQGNKAEKAGSKEQGPRQRKGAAPAEKKCGAETQHEGLELRVENLQAVQTDFSSDPLQKVVCFNHDNTLLATGGTDGYVRVWKVPSLEKVLEFKAHEGEIEDLALGPDGKLVTVGRDLKASVWQKDQLVTQLHWQENGPT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PREB(10113)

General description

The gene PREB (prolactin regulatory element binding) is also referred to as Sec12 and is mapped to human chromosome 2p23. The encoded 417 amino acids protein contains highly conserved seven WD-40 folds. It is localized to the ER (endoplasmic reticulum) exit sites in mammalian cells. The encoded protein contains two PQ-rich potential transactivation domains and lacks a DNA binding motif. This gene overlaps the αβhydrolase domain containing 1 (ABHD1) gene along a stretch of 42bp and may be associated with some of the developmental abnormalities found in patients with partial trisomy 2p.

Immunogen

Prolactin regulatory element-binding protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The gene PREB (prolactin regulatory element binding) encodes a type II transmembrane protein. It binds to the Pit1-binding element of the prolactin (PRL) promoter and plays a transcriptional regulatory role in the pituitary and other organs in which the protein is expressed. In response to cytokines, PREB transcriptionally regulates MCP-1 (monocyte chemoattractant protein-1) promoter. MCP-1 functions as an initiating factor in the inflammatory process of atherogenesis. PREB also functions as a guanine-nucleotide exchange factor for Sar1 guanosine triphosphatase and is essential for collagen VII secretion. It is recruited to ER (endoplasmic reticulum) exit sites by its interaction with a collagen cargo receptor component, cTAGE5. The export of large cargo at the ER exit sites involves high levels of guanosine triphosphate-bound Sar1 generated by PREB.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72889

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kota Saito et al.
The Journal of cell biology, 206(6), 751-762 (2014-09-10)
Mechanisms for exporting variably sized cargo from the endoplasmic reticulum (ER) using the same machinery remain poorly understood. COPII-coated vesicles, which transport secretory proteins from the ER to the Golgi apparatus, are typically 60-90 nm in diameter. However, collagen, which
Koji Murao et al.
Atherosclerosis, 207(1), 45-50 (2009-05-12)
The prolactin regulatory element binding (PREB) protein is a transcriptional factor that regulates prolactin promoter activity in rat anterior pituitary. It is expressed not only in the anterior pituitary but also in the cardiovascular system, including in human umbilical vascular
M S Fliss et al.
Molecular endocrinology (Baltimore, Md.), 13(4), 644-657 (1999-04-09)
Previous studies have implied that a transcription factor(s) other than Pit-1 is involved in homeostatic regulation of PRL promoter activity via Pit-1-binding elements. One such element, 1P, was employed to clone from a rat pituitary cDNA expression library a novel
C L Taylor Clelland et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 11(8), 675-681 (2000-08-02)
We have isolated the human homolog of a novel rodent gene that may be involved in the regulation of pituitary gene transcription. The human PREB gene encodes a predicted protein of 417 amino acids, exhibiting several sequences characteristic of the
Alasdair J Edgar
BMC genomics, 4(1), 18-18 (2003-05-09)
Overlapping sense/antisense genes orientated in a tail-to-tail manner, often involving only the 3'UTRs, form the majority of gene pairs in mammalian genomes and can lead to the formation of double-stranded RNA that triggers the destruction of homologous mRNAs. Overlapping polyadenylation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service