Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA013355

Sigma-Aldrich

Anti-P3H2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ10718, Anti-LEPREL1, Anti-MLAT4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

ANPEHMEMQQNIENYRATAGVEALQLVDREAKPHMESYNAGVKHYEADDFEMAIRHFEQALREYFVEDTECRTLCEGPQRFEEYEYLGYKAGLYEAIADHYMQVLVCQHECV

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LEPREL1(55214)

General description

LEPREL1 (leprecan-like 1) gene is also called P3H2 (prolyl 3-hydroxylase 2) and encodes an enzyme that belongs to the prolyl 3-hydroxylase family. It is expressed in placenta, lung, heart, and kidney. The encoded protein has a molar mass of 80kDa with 708 amino acids and is expressed as a 3.4kb transcript. The protein contains a signal sequence, four tetratricopeptide repeats (TPRs), a leucine zipper, a P-loop, a prolyl 4-hydroxylase α domain (P4Hα), and a C-terminal KDEL ER-retention motif. It is localized to the ER and Golgi network.

Immunogen

prolyl 3-hydroxylase 2

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LEPREL1 (leprecan-like 1) gene encodes prolyl 3-hydroxylase 2 (P3H2), which is a 2-oxoglutarate-dependent dioxygenase that hydroxylates collagens. It catalyzes the post-translational formation of 3-hydroxyproline in -Xaa-Pro-Gly-sequences in types II, IV and V collagens. It is involved in the post-translational modification of fibril-forming collagens. Mutation in this gene is associated with high-grade axial myopia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71294

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sofia Järnum et al.
Biochemical and biophysical research communications, 317(2), 342-351 (2004-04-06)
We have identified a novel protein, Leprecan-like 1 (LEPREL1), with profound similarity to the Leprecan family of proteoglycans. The genomic organization of the Leprecan gene family was found to be highly conserved. Expression analysis shows that LEPREL1 is expressed in
Shikma Mordechai et al.
American journal of human genetics, 89(3), 438-445 (2011-09-03)
Autosomal-recessive high-grade axial myopia was diagnosed in Bedouin Israeli consanguineous kindred. Some affected individuals also had variable expressivity of early-onset cataracts, peripheral vitreo-retinal degeneration, and secondary sight loss due to severe retinal detachments. Through genome-wide linkage analysis, the disease-associated gene
Russell J Fernandes et al.
The Journal of biological chemistry, 286(35), 30662-30669 (2011-07-16)
The fibrillar collagen types I, II, and V/XI have recently been shown to have partially 3-hydroxylated proline (3Hyp) residues at sites other than the established primary Pro-986 site in the collagen triple helical domain. These sites showed tissue specificity in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service