Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA012782

Sigma-Aldrich

Anti-ROMO1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MGR2 protein homolog, Anti-ROS modulator 1, AntiReactive oxygen species modulator 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

GRELMGGIGKTMMQSGGTFGTFMAIGMGIRC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ROMO1(140823)

General description

Reactive oxygen species modulator 1 (ROMO1) is a transmembrane protein localized in the mitochondria.

Immunogen

Reactive oxygen species modulator 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Reactive oxygen species modulator 1 (ROMO1) functions as an antibacterial polypeptide. It can act against a variety of bacteria like multidrug resistant (MDR) tubercle bacillus, methicillin-resistant Staphylococcus aureus and MDR Pseudomonas aeruginosa. ROMO1 causes premature senescence by reactive oxygen species production and this further leads to the induction of DNA damage. Knockdown of the protein inhibits the progression of replicative senescence, leads to cell cycle arrest at the G1 phase and inhibits cyclin D1 expression. ROMO1 triggers S-phase kinase-associated protein 2 (Skp2)-mediated ubiquitylation and degradation of Myc (a transcription factor) in a negative-feedback mechanism.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71895.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antibacterial potential of hGlyrichin encoded by a human gene.
Sha J, et al.
Journal of Peptide Science, 18(2), 97-104 (2012)
Romo1 is a negative-feedback regulator of Myc.
Lee SB, et al.
Journal of Cell Science, 124(Pt 11), 1911-1924 (2011)
Replicative senescence induced by Romo1-derived reactive oxygen species.
Chung YM, et al.
The Journal of Biological Chemistry, 283(48), 33763-33771 (2008)
Miha Tibaut et al.
BMC medical genomics, 13(1), 184-184 (2020-12-12)
We aimed to examine the role of the rs6060566 polymorphism of the reactive oxygen species modulator 1 (ROMO1) gene in the development of myocardial infarction (MI) in Caucasians with type 2 diabetes (T2DM). A total of 1072 subjects with T2DM

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service