Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA012638

Sigma-Aldrich

Anti-GALNT8 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GalNAc-T8, Anti-Polypeptide GalNAc transferase 8, Anti-Probable polypeptide N-acetylgalactosaminyltransferase 8, Anti-Protein-UDP acetylgalactosaminyltransferase 8, Anti-UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 8, Anti-pp-GaNTase 8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

NLLDENVCLDQGPFPGNTPIMYYCHEFSSQNVYYHLTGELYVGQLIAEASASDRCLTDPGKAEKPTLEPCSKAAKNRLHIYWDFKPGGAVINRDTKRCLEMKKDLLGSHVLVLQTCSTQVWEIQHTVR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GALNT8(26290)

General description

Polypeptide GalNAc transferase 8 (GALNT8) is a type-II membrane protein which belongs to the UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase (ppGaNTase) family. The gene is present on chromosome 12p13.3 and encodes 637 amino acids.

Immunogen

polypeptide N-acetylgalactosaminyltransferase 8

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Polypeptide GalNAc transferase 8 (GALNT8) initiates mucin-like O-glycosylation of specific target proteins and transfers GalNAc to serine and threonine residues on target proteins in the Golgi apparatus. Hence it takes part in the biosynthesis of mucin-like O-glycans. It may modulate the signal transduction of IFN (Interferon)-α-mediated antiviral activity. The overexpression of GALNT8 weakens the IFN-α-induced gene transcription via the IFN-stimulated response element. Thus, it is linked with the response to IFN therapy against chronic hepatitis C (CHC).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71913

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-nucleotide polymorphisms in GALNT8 are associated with the response to interferon therapy for chronic hepatitis C.
R Nakano
The Journal of General Virology, 94(Pt 1), 81-89 (2013)
K E White et al.
Gene, 246(1-2), 347-356 (2000-04-18)
The UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase (ppGaNTase) family of enzymes initiates mucin-like O-glycosylation of specific proteins. Using exon-prediction analysis on genomic sequence from human chromosome 12p13.3, we identified novel exons that shared significant homology with the ppGaNTases. cDNA library screening and RT-PCR produced
Peiwei Chai et al.
Nucleic acids research, 46(12), 6041-6056 (2018-05-10)
Aberrant chromatin transformation dysregulates gene expression and may be an important driver of tumorigenesis. However, the functional role of chromosomal dynamics in tumorigenesis remains to be elucidated. Here, using in vitro and in vivo experiments, we reveal a novel long

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service