Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA012585

Sigma-Aldrich

Anti-SCN2B antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Sodium channel subunit beta-2 precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

KEQKLSTDDLKTEEEGKTDGEGNPDDGAK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SCN2B(6327)

Immunogen

Sodium channel subunit beta-2 precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-SCN2B antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SCN2B (Sodium channel, voltage-gated, type II, β subunit) is a 33kDa transmembrane type II glycoprotein belonging to the the IgCAM superfamily. It consists of a single transmembrane domain and an immunoglobulin-like fold at the extracellular end. It plays an important role in controlling the cell-surface β2 expression, cell adhesion-migration, and voltage-gated sodium channeling. Missense mutation in SCN2B function is associated with Brugada syndrome, and atrial fibrillation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72061.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helena Riuró et al.
Human mutation, 34(7), 961-966 (2013-04-06)
Brugada Syndrome (BrS) is a familial disease associated with sudden cardiac death. A 20%-25% of BrS patients carry genetic defects that cause loss-of-function of the voltage-gated cardiac sodium channel. Thus, 70%-75% of patients remain without a genetic diagnosis. In this
Doo Yeon Kim et al.
The Journal of biological chemistry, 280(24), 23251-23261 (2005-04-19)
The voltage-gated sodium channel beta2-subunit (beta2) is a member of the IgCAM superfamily and serves as both an adhesion molecule and an auxiliary subunit of the voltage-gated sodium channel. Here we found that beta2 undergoes ectodomain shedding followed by presenilin
Hiroshi Watanabe et al.
Circulation. Arrhythmia and electrophysiology, 2(3), 268-275 (2009-10-08)
We and others have reported mutations in the cardiac predominant sodium channel gene SCN5A in patients with atrial fibrillation (AF). We also have reported that SCN1B is associated with Brugada syndrome and isolated cardiac conduction disease. We tested the hypothesis
J Eubanks et al.
Neuroreport, 8(12), 2775-2779 (1997-08-18)
The beta2 subunit of rat brain voltage-gated sodium channels modulates their cell-surface expression and gating. It is a 33 kDa glycoprotein with a single transmembrane segment and an immunoglobulin-like fold resembling those of cell adhesion molecules in the extracellular domain.
Diana Shy et al.
Circulation, 130(2), 147-160 (2014-06-05)
Sodium channel NaV1.5 underlies cardiac excitability and conduction. The last 3 residues of NaV1.5 (Ser-Ile-Val) constitute a PDZ domain-binding motif that interacts with PDZ proteins such as syntrophins and SAP97 at different locations within the cardiomyocyte, thus defining distinct pools

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service