Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA012130

Sigma-Aldrich

Anti-ADAM23 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ADAM metallopeptidase domain 23, Anti-MDC3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LLVLLLLPPLAASSRPRAWGAAAPSAPHWNETAEKNLGVLADEDNTLQQNSSSNISYSNAMQKEITLPSRLIYYINQDSESPYHVLDTKARHQQKHNKAVHLAQASFQIEAFGSKFILD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ADAM23(8745)

Immunogen

ADAM metallopeptidase domain 23 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ADAM23 (ADAM metallopeptidase domain 23) is a cell surface protein expressed in normal stomach tissues. It is involved in the cell-cell and/or cell-matrix interactions and protease activities. It also possesses tumor suppression property. It has been reported that methylated ADAM23 shows downregulated expression in the brain tumors and it may explain the cause of brain tumor metastasis. It has also been reported that any functional alteration of ADAM23 through hypermethylated genetic or epigenetic silencing may lead to gastric carcinogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71834.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisa Helena Farias Jandrey et al.
Neuro-oncology advances, 5(1), vdad147-vdad147 (2023-11-29)
Infiltration is a life-threatening growth pattern in malignant astrocytomas and a significant cause of therapy resistance. It results in the tumor cell spreading deeply into the surrounding brain tissue, fostering tumor recurrence and making complete surgical resection impossible. We need
Hisashi Takada et al.
Oncogene, 24(54), 8051-8060 (2005-08-17)
Array-based comparative genomic hybridization (CGH-array) has a powerful potential for high-throughput identification of genetic aberrations in cell genomes. We identified a homozygous loss of ADAM23 (2q33.3) in the course of a program to screen a panel of gastric cancer (GC)
Fabrício F Costa et al.
Neuroscience letters, 380(3), 260-264 (2005-05-03)
The ADAMs comprises a family of cell surface proteins with putative roles in cell-cell and/or cell-matrix interactions and in protease activities. In this work, we have examined the expression level and the methylation status of the 5' upstream region of
E T Costa et al.
Oncogene, 34(10), 1270-1279 (2014-03-26)
Intratumoral heterogeneity (ITH) represents an obstacle for cancer diagnosis and treatment, but little is known about its functional role in cancer progression. The A Desintegrin And Metalloproteinase 23 (ADAM23) gene is epigenetically silenced in different types of tumors, and silencing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service