Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA011960

Sigma-Aldrich

Anti-CAPN15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C11orf69, Anti-CAPN15, Anti-G2, Anti-MGC34830, Anti-SOLH, Anti-small optic lobes homolog (Drosophila)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

FLPVLNGVLPKPPAILGEPKGSCQEEAGPVRTAGLVATEPARGQCEDKDEEEKEEQEEEEGAAEPRGGWACPRCTLHNTPVASSCSVCGGPRRLSLPRIPPEALVVPE

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOLH(6650)

Immunogen

small optic lobes homolog (Drosophila) recombinant protein epitope signature tag (PrEST)

application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CAPN15 (Calpain 15) is a member of the calpain and zinc-finger gene family consisting of a zinc-finger-like domain, a conserved calpain-like protease domain, and a C-terminal region. It is expressed in human brain. This cytoplasmic cysteine protease is functionally activated by the presence of calcium. Calpain is involved in various calcium-regulated cellular processes such as signal transduction, cell proliferation, cell cycle progression, differentiation, apoptosis, membrane fusion, and platelet activation. SOLH is a candidate gene for CATM (hereditary cataracts with microphthalmia). It also participates in the catabolic digestion of protein in diet composition and/or energy restriction.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70874

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A N Belcastro et al.
Canadian journal of applied physiology = Revue canadienne de physiologie appliquee, 21(5), 328-346 (1996-10-01)
Although the proteolytic events accompanying acute and chronic perturbations in striated muscle protein turnover remain to be fully elucidated, the purpose of this paper is to (a) review the chemistry of the nonlysosomal calpain-calpastatin system, and (b) provide evidence for
Koichi Suzuki et al.
Diabetes, 53 Suppl 1, S12-S18 (2004-01-30)
Variation in the calpain 10 gene has recently been shown to be associated with type 2 diabetes by positional cloning. Since then, studies on calpain 10 have been started in correlation with diabetes and insulin-mediated signaling. In this review, the
M Kamei et al.
Genomics, 51(2), 197-206 (1998-09-02)
Mutations in the Drosophila melanogaster small optic lobes (sol) gene cause a sever reduction in the neuropiles of the medulla and lobula complexes of the adult optic lobes. The predicted protein product of sol contains zinc-finger-like repeats, a calpain-like protease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service