Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA011356

Sigma-Aldrich

Anti-PCGF1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-NSPc1, Anti-Nervous system Polycomb-1, Anti-Polycomb group RING finger protein 1, Anti-RING finger protein 68

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LVPGLQDSEEKRIREFYQSRGLDRVTQPTGEEPALSNLGLPFSSFDHSKAHYYRYDEQLNLCLERLSSGKDKNKSVLQNKYVRCSVRAEVRHLRRVLCHRLMLNPQHVQLLFDNEVLPDHMTMKQIWLSRWFGKPSPLLLQYSVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCGF1(84759)

General description

PCGF1 (polycomb group ring finger 1) is a member of the polycomb group (PcG) proteins. PcG in mammals, are divided into two polycomb repressive complexes (PRCs), and PCGF1 belongs to PRC1. It shares high homology to the other two members of PRC1 namely, Mel-18 and Bmi-1. This protein predominantly resides in the nucleus. It has a RING finger domain in its N-terminal, and is ubiquitously expressed in adult tissues. This gene maps to human chromosome 2p13, and spans around 400kb.

Immunogen

Polycomb group RING finger protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PCGF1 (polycomb group ring finger 1) is involved in the repression of transcription, and is a biomarker for kidney cancer. Studies show that it promotes tumorigenesis and cell proliferation by repressing the expression of p21 through RARE (retinoid acid response element). This gene is located in the region linked to susceptibility to Parkinson′s disease (PD), and therefore, it might be involved in the functional and pathological processes related to adult nervous system. In mouse, this gene is located with the locus of lethal neuromuscular wasting disorder (mnd2). It is also highly expressed in the developing nervous system, predominantly in the neural crest cells, which give rise to peripheral nervous system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71061

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanhua Gong et al.
FEBS letters, 579(1), 115-121 (2004-12-29)
Nervous system polycomb 1 (NSPc1) shares high homology with vertebrate PcG proteins Mel-18 and Bmi-1. The mRNA of NSPc1 is highly expressed in the developmental nervous system [Mech. Dev. 102 (2001) 219-222]. However, the functional characterization of NSPc1 protein is
Yanhua Gong et al.
Nucleic acids research, 34(21), 6158-6169 (2006-11-08)
The mammalian polycomb group proteins play an important role in cell cycle control and tumorigenesis. Nervous system polycomb 1 (NSPc1) is a newly identified transcription repressor, highly homologous with PcG protein Bmi-1. In this article, we showed that NSPc1 could
M Nunes et al.
Mechanisms of development, 102(1-2), 219-222 (2001-04-05)
Anterior-posterior (A-P) patterning is a key element in early embryonic development. Polycomb group (PcG) genes act as transcriptional repressors to regulate A-P patterning by either directly or indirectly controlling the coordinated expression of the HOM/Hox homeobox (Curr. Opin. Genet. Dev.
Gergely Rona et al.
eLife, 7 (2018-07-10)
The mammalian FBXL10-RNF68-RNF2 ubiquitin ligase complex (FRRUC) mono-ubiquitylates H2A at Lys119 to repress transcription in unstressed cells. We found that the FRRUC is rapidly and transiently recruited to sites of DNA damage in a PARP1- and TIMELESS-dependent manner to promote

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service