Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA011129

Sigma-Aldrich

Anti-NDST4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 4 antibody produced in rabbit, Anti-Glucosaminyl N-deacetylase/N-sulfotransferase 4 antibody produced in rabbit, Anti-N-HSST 4 antibody produced in rabbit, Anti-N-heparan sulfate sulfotransferase 4 antibody produced in rabbit, Anti-NDST-4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

GYKQEMTLIETTAEAECTDIKILPYRSMELKTVKPIDTSKTDPTVLLFVESQYSQLGQDIIAILESSRFQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDST4(64579)

General description

NDST4 (N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 4) is the fourth member of NDST family, and shows high sequence homology to the other three members namely, NDST1, NDST2 and NDST3. It is made of 872 amino acids. This gene is localized to human chromosome 4q26, and alternative splicing gives rise to two isoforms of this protein. This protein is majorly expressed in developing embryos, and in adult brain. It is a type II transmembrane protein with 22 amino acids in the transmembrane region, and 12-13 amino acids in the cytoplasmic domain. It also shares two PAPS (3′-phosphoadenosine 5′-phosphosulfate) binding motifs, with other sulfotransferases.

Immunogen

Bifunctional heparan sulfate N-deacetylase/N-sulfotransferase 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The functionality of NDST4 (N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 4) is not yet characterized. It has higher sulfotransferase activity and lower deacetylase activity. It catalyzes the synthesis of heparan sulfate (HS) on existing proteins to form heparan sulfate proteoglycans (HSPGs). This gene is down-regulated in colorectal cancer (CRC), and is associated with advanced cancer phenotype. It is associated with invasiveness and metastasis of CRC, which might be an outcome of alterations in the HS chains of certain HSPGs. Therefore, NDST4 might have potential as a marker to predict the prognosis of CRC. This protein might be responsible for the addition of sulfate groups to glucosaminyl residues, which were deacetylated and left unaltered by other NDST members. Genome-wide association studies show that SNP near this gene is associated with maximum number of drinks (MaxDrinks), which is related to alcohol dependence.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71993

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng-Tai Tzeng et al.
PloS one, 8(6), e67040-e67040 (2013-07-05)
Genomic deletion at tumor suppressor loci is a common genetic aberration in human cancers. The study aimed to explore candidate tumor suppressor genes at chromosome 4q25-q28.2 and to delineate novel prognostic biomarkers associated with colorectal cancer (CRC). Deletion mapping of
Yue Pan et al.
Journal of psychiatric research, 47(11), 1717-1724 (2013-08-21)
Maximum number of drinks (MaxDrinks) defined as "Maximum number of alcoholic drinks consumed in a 24-h period" is an intermediate phenotype that is closely related to alcohol dependence (AD). Family, twin and adoption studies have shown that the heritability of
J Aikawa et al.
The Journal of biological chemistry, 276(8), 5876-5882 (2000-11-23)
We report the cloning and partial characterization of the fourth member of the vertebrate heparan sulfate/heparin: GlcNAc N-deacetylase/GlcN N-sulfotransferase family, which we designate NDST4. Full-length cDNA clones containing the entire coding region of 872 amino acids were obtained from human
Kay Grobe et al.
The Journal of biological chemistry, 277(34), 30699-30706 (2002-06-19)
We report the full-length 5'-untranslated region (5'-UTR) sequences of the four vertebrate heparan sulfate/heparin GlcNAc N-deacetylase/N-sulfotransferases (NDSTs) and their role in translational regulation in vivo and in vitro. All four NDST 5'-UTR sequences are unusually long, have a high degree

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service