Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA011046

Sigma-Aldrich

Anti-MANEA antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Endo-α mannosidase, Anti-Endomannosidase, Anti-Glycoprotein endo-α-1,2-mannosidase, Anti-Mandaselin, Anti-hEndo

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

LRPNTATFGAPFGLDLLPELHQRTIHLGKNFDFQKSDRINSETNTKNLKSVEITMKPSKASELNLDELPPLNNYLHVFYYSWYGNPQFDGKYIHWNHPVLEHWDPRIAKNYPQGRHNPPDDIGSSFYPELGSYSSRD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MANEA(79694)

Looking for similar products? Visit Product Comparison Guide

General description

MANEA (mannosidase, endo-α) is a type II transmembrane glycosidic protein. This enzyme is made of 462 amino acids and has a molecular weight of 53.6kDa. Its transmembrane region is made of 19 amino acids and lies in close proximity to the N-terminal. This gene is present on human chromosome 6q16.1, and is expressed in multiple human tissues including brain, kidney and liver.

Immunogen

Glycoprotein endo-α-1,2-mannosidase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MANEA (mannosidase, endo-α) catalyzes the cleavage of mono-, di-, and tri-glucosyl-mannose oligosaccharides from high-mannose N-glycans, by breaking the α-1,2-mannosidic bond. Studies also suggest that MANEA is involved in ensuring proper N-glycosylation, and thus provides cells with the ability to recover and mature glycosylated structures. Single nucleotide polymorphisms (SNPs) in this gene are linked to cocaine dependence and cocaine induced paranoia in European American and African American populations. Studies suggest that variations in MANEA gene are associated with anxiety disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72259

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolay V Kukushkin et al.
The Biochemical journal, 438(1), 133-142 (2011-05-19)
During quality control in the ER (endoplasmic reticulum), nascent glycoproteins are deglucosylated by ER glucosidases I and II. In the post-ER compartments, glycoprotein endo-α-mannosidase provides an alternative route for deglucosylation. Previous evidence suggests that endomannosidase non-selectively deglucosylates glycoproteins that escape
K P Jensen et al.
Translational psychiatry, 4, e353-e353 (2014-01-30)
Unbiased genome-wide approaches can provide novel insights into the biological pathways that are important for human behavior and psychiatric disorder risk. The association of α-endomannosidase gene (MANEA) variants and cocaine-induced paranoia (CIP) was initially described in a study that used
Lindsay A Farrer et al.
Archives of general psychiatry, 66(3), 267-274 (2009-03-04)
Cocaine dependence (CD) and related behaviors are highly heritable, but no genetic association has been consistently demonstrated. A recent genome-wide study of drug dependence identified an association between cocaine-induced paranoia (CIP) and a single-nucleotide polymorphism (SNP) in the alpha-endomannosidase (MANEA)
Stephen R Hamilton et al.
Glycobiology, 15(6), 615-624 (2005-01-29)
Rat endomannosidase is a glycosidic enzyme that catalyzes the cleavage of di-, tri-, or tetrasaccharides (Glc(1-3)Man), from N-glycosylation intermediates with terminal glucose residues. To date it is the only characterized member of this class of endomannosidic enzymes. Although this protein
Birgit Hardt et al.
Biochimie, 87(2), 169-179 (2005-03-12)
The cDNA for human endo-alpha1,2-mannosidase was reconstructed using two independent EST-clones and its properties characterized. The 2837 bp cDNA construct contained a 1389 bp open reading frame (ORF) encoding for 462 amino acids and an approximately 53.6 kDa protein, respectively.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service