Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA010972

Sigma-Aldrich

Anti-VMA21 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AF003627.1, Anti-UPF0569 transmembrane protein LOC203547

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PRRTMLRGKSRLNVEWLGYSPGLLLEHRPLLAGRTPRSHRR

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VMA21(203547)

General description

VMA21 (vacuolar membrane ATPase activity 21 (VMA21)) is a diverged ortholog of yeast Vma21p protein, which is found in humans. VMA21 forms a part of the vacuolar ATPases (V-ATPases), which are expressed in the endomembrane systems of all eukaryotes. It has 14 subunits arranged in two domains, transmembrane V0 domain and cytoplasmic V1 domain. In humans, this gene is localized to chromosome Xq28.

Immunogen

UPF0569 transmembrane protein LOC203547 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

VMA21 (vacuolar membrane ATPase activity 21 (VMA21)) initiates the assembly of the V0 domain, which acts as a proton-translocator, of vacuolar ATPases (V-ATPases). It interacts with the c′′ subunit of V0 domain. It transports this domain from endoplasmic reticulum to Golgi bodies, in COPII vesicles. V-ATPases are required to maintain the acidity of lysosomes, as well as the pH gradient of the endovesicular secretory pathways. Therefore, inactivation of VMA21 leads to an increase in the lysosomal pH. Mutations in this gene lead to X-linked myopathy with excessive autophagy (XMEA), which is characterized by vacuolation and atrophy of skeletal muscles.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74403

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fine-mapping the gene for X-linked myopathy with excessive autophagy.
I Munteanu et al.
Neurology, 71(12), 951-953 (2008-09-17)
Sally Esmail et al.
The Journal of biological chemistry, 293(8), 2787-2800 (2018-01-10)
The a subunit is the largest of 15 different subunits that make up the vacuolar H+-ATPase (V-ATPase) complex, where it functions in proton translocation. In mammals, this subunit has four paralogous isoforms, a1-a4, which may encode signals for targeting assembled
Sally Esmail et al.
Journal of cellular biochemistry, 119(1), 861-875 (2017-07-01)
The a subunit of the V
Michio Hirano et al.
Cell, 137(2), 213-215 (2009-04-22)
The Vma21p protein in yeast is an essential assembly chaperone for the vacuolar ATPase, the major proton pump of cellular membranes. In this issue, Ramachandran et al. (2009) report that mutations in the gene encoding the human homolog VMA21 cause
Nivetha Ramachandran et al.
Acta neuropathologica, 125(3), 439-457 (2013-01-15)
X-linked Myopathy with Excessive Autophagy (XMEA) is a childhood onset disease characterized by progressive vacuolation and atrophy of skeletal muscle. We show that XMEA is caused by hypomorphic alleles of the VMA21 gene, that VMA21 is the diverged human ortholog

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service