Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA009668

Sigma-Aldrich

Anti-SERPINB6 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CAP antibody produced in rabbit, Anti-Cytoplasmic antiproteinase antibody produced in rabbit, Anti-PI-6 antibody produced in rabbit, Anti-Placental thrombin inhibitor antibody produced in rabbit, Anti-Protease inhibitor 6 antibody produced in rabbit, Anti-Serpin B6 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

FALNLLKTLGKDNSKNVFFSPMSMSCALAMVYMGAKGNTAAQMAQILSFNKSGGGGDIHQGFQSLLTEVNKTGTQYLLRMANRLFGEKSCDFLSSFRDSCQKFYQAEMEELDFISAVEKSRKHINTWVAEKTEGKIAELLSPGSV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINB6(5269)

Looking for similar products? Visit Product Comparison Guide

General description

SERPINB6 (serpin peptidase inhibitor, clade B, member 6), also called cytoplasmic antiproteinase (CAP), is an intracellular serpin, which shows a wide range of tissue expression including platelets. It is a 70kDa member of the ovalbumin family of serpins.

Immunogen

Serpin B6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SERPINB6 (serpin peptidase inhibitor, clade B, member 6) is responsible for quick and efficient neutralization of chymotrypsin. The expression of this protein is linked with platelet development. It inhibits the activity of multiple proteases such as thrombin, urokinase, and factor Xa. Mutation in this gene resulting in truncated protein formation is linked with autosomal-recessive nonsyndromic sensorineural hearing loss. It is candidate marker for colon cancer, where its expression is lost.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70761

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G S Krasnov et al.
Molekuliarnaia biologiia, 43(2), 348-356 (2009-05-12)
Modern proteomic techniques make it possible to identify numerous changes in protein expression in tumor in comparison to normal tissues. Despite the wide application of proteomics in current studies, identification of proteins with stable concentration differences in normal and cancer
M Riewald et al.
The Journal of biological chemistry, 271(24), 14526-14532 (1996-06-14)
Human cytoplasmic antiproteinase (CAP) is an intracellular serpin that has been reported to utilize Arg341 as the reactive site P1 residue to neutralize a broad variety of extracellular serine proteases with trypsin-like specificity. Both native CAP and recombinant CAP purified
M Riewald et al.
The Journal of biological chemistry, 271(12), 7160-7167 (1996-03-22)
To define the presence and potential role of platelet-associated protease inhibitors, we initiated a study designed to characterize the platelet components that are responsible for the formation of two SDS-stable complexes of approximately 58 and 70 kDa initially observed following
Asli Sirmaci et al.
American journal of human genetics, 86(5), 797-804 (2010-05-11)
More than 270 million people worldwide have hearing loss that affects normal communication. Although astonishing progress has been made in the identification of more than 50 genes for deafness during the past decade, the majority of deafness genes are yet

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service