Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA009411

Sigma-Aldrich

Anti-IMPAD1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ20421, Anti-IMPA3, Anti-gPAPP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

NVLHEKSKGKTREGAEDKMTSGDVLSNRKMFYLLKTAFPSVQINTEEHVDAADQEVILWDHKIPEDILKEVTTPKEVPAESVTVWIDPLDATQEYTEDLRKYVTTMVCVAVNGK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IMPAD1(54928)

General description

IMPAD1 (inositol monophosphatase domain containing 1) is a nucleotide phosphatase which is localized to Golgi, and hence, is also called Golgi-resident PAP phosphatase (gPAPP). This gene is localized to human chromosome 8. This protein was first identified in metazoan genome, and is composed of a single transmembrane domain. It is a type II transmembrane protein.

Immunogen

myo-inositol monophosphatase A3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In mice, IMPAD1 (inositol monophosphatase domain containing 1) inactivation is linked with skeletal dysplasia and abnormal joint formation. It hydrolyzes the byproduct of sulfotransferase ractions, phosphoadenosine phosphate (PAP) to AMP. In human, the deficiency of this enzyme is linked with chondrodysplasia, which is a skeletoarticular disorder linked with defective synthesis of sulfated proteoglycans. Mutations in this gene are linked with short stature, joint dislocations, brachydactyly and cleft palate in patients with Desbuquois dysplasia type 1. Mutations in IMPAD1 are also associated with Catel-Manzke syndrome, the patients of which show hyperphalangism with index fingers demonstrating bilateral deviation, and micrognathia with or without cleft palate. In vitro this protein is inhibited by lithium, and mice with IMPAD1 inactivation show neonatal fatality lung aberrations similar to atelectasis, and dwarfism characterized by abnoraml cartilage structure. This protein might be involved in endochondral ossification, thus, playing a part in formation of skeletal elements.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71722

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rakhee Bajaj et al.
Cell reports, 40(13), 111429-111429 (2022-09-29)
Lung cancer is a highly aggressive and metastatic disease responsible for approximately 25% of all cancer-related deaths in the United States. Using high-throughput in vitro and in vivo screens, we have previously established Impad1 as a driver of lung cancer invasion and
Joshua P Frederick et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(33), 11605-11612 (2008-08-13)
Sulfation is an important biological process that modulates the function of numerous molecules. It is directly mediated by cytosolic and Golgi sulfotransferases, which use 3'-phosphoadenosine 5'-phosphosulfate to produce sulfated acceptors and 3'-phosphoadenosine 5'-phosphate (PAP). Here, we identify a Golgi-resident PAP
Lisenka E L M Vissers et al.
American journal of human genetics, 88(5), 608-615 (2011-05-10)
We used whole-exome sequencing to study three individuals with a distinct condition characterized by short stature, chondrodysplasia with brachydactyly, congenital joint dislocations, cleft palate, and facial dysmorphism. Affected individuals carried homozygous missense mutations in IMPAD1, the gene coding for gPAPP
Mathilde Nizon et al.
American journal of medical genetics. Part A, 158A(9), 2183-2187 (2012-08-14)
Catel-Manzke syndrome is characterized by hyperphalangism with bilateral deviation of the index fingers and micrognathia with or without cleft palate. Some atypical patients present with additional malformations. No molecular basis is yet available. Most patients have an unremarkable family history

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service