Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA008835

Sigma-Aldrich

Anti-CPT1A antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CPT I, Anti-CPTI-L, Anti-Carnitine O-palmitoyltransferase I, liver isoform, Anti-Carnitine palmitoyltransferase 1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.4 μg/mL
immunofluorescence: 1-4 μg/mL
immunohistochemistry: 1:50- 1:200

immunogen sequence

ARCRQAYFGRGKNKQSLDAVEKAAFFVTLDETEEGYRSEDPDTSMDSYAKSLLHGRCYDRWFDKSFTFVVFKNGKMGLNAEHSWADAPIVAHLWEYVMSIDSLQLGYA

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CPT1A(1374)

General description

CPT1A (carnitine palmitoyltransferase 1A) is a mitochondrial outer membrane protein, which is transcribed into three isoforms, namely CPT1A, CPT1B and CPT1C. CPT1A is widely expressed with a predominant expression in liver, CPT1B is abundant in muscle and CPT1C is majorly found in the brain.[1] CPT1A is also found to be expressed on CD4+ T-cells.

Immunogen

Carnitine O-palmitoyltransferase 1, liver isoform recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CPT1A (carnitine palmitoyltransferase 1A) enzyme catalyzes the synthesis of long chain acyl carnitine. This then crosses the inner mitochondrial membrane and finally is subjected to β-oxidation in the mitochondria. It is the major enzyme involved in the regulation of fatty acid oxidation, and has potential as a therapeutic target for the treatment of metabolic syndrome. It is thought to play a role in cancer and apoptosis, and in neoplastic cells, CPT1A resides in the nucleus and interacts with histone deacetylase complex.[1] The inactivation of this enzyme is linked with oxidative abnormality in mitochondrial metabolism. This is usually a result of stress conditions resulting in the generation of reactive oxygen species (ROS). Polymorphism CPT1A p.P479L is common to certain coastal British Columbia First Nations, and is linked with sudden death in infants.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70469.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marguerite R Irvin et al.
Circulation, 130(7), 565-572 (2014-06-13)
Genetic research regarding blood lipids has largely focused on DNA sequence variation; few studies have explored epigenetic effects. Genome-wide surveys of DNA methylation may uncover epigenetic factors influencing lipid metabolism. To identify whether differential methylation of cytosine-(phosphate)-guanine dinucleotides (CpGs) correlated
Daiki Setoyama et al.
Genes to cells : devoted to molecular & cellular mechanisms, 18(12), 1107-1119 (2013-10-15)
Oxidative dysfunction in the metabolism has long been implicated in diverse biological disorders. Although a substantial number of metabolic enzymes are targeted for inactivation by oxidative stress, identifying those targets remains difficult due to a lack of comprehensive observations of
Graham B Sinclair et al.
Pediatrics, 130(5), e1162-e1169 (2012-10-24)
Infant mortality in British Columbia (BC) First Nations remains elevated relative to other residents. The p.P479L (c.1436C>T) variant of carnitine palmitoyltransferase 1 (CPT1A) is frequent in some aboriginal populations and may be associated with increased infant deaths. This work was
Lingling Liu et al.
BMC cancer, 12, 154-154 (2012-04-27)
Alveolar rhabdomyosarcoma (ARMS) has a high propensity to metastasize, leading to its aggressiveness and a poor survival rate among those with the disease. More than 80% of aggressive ARMSs harbor a PAX3-FKHR fusion transcription factor, which regulates cell migration and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service