Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA008174

Sigma-Aldrich

Anti-SHANK2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CortBP1, Anti-Cortactin-binding protein 1, Anti-SH3 and multiple ankyrin repeat domains protein 2, Anti-Shank2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SELNSILQQMNREKLAKPGEGLDSPMGAKSASLAPRSPEIMSTISGTRSTTVTFTVRPGTSQPITLQSRPPDYESRTSGTRRAPSPVVSPTEMNKETLPAPLSAATASPSPALSDVFSLPSQPPSGDLFG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SHANK2(22941)

General description

SHANK2 (SH3 and multiple ankyrin repeat domains 2) gene encodes a scaffold protein belonging to the Shank family of synaptic proteins. The Shank proteins contain multiple ankyrin repeats, an SH3 domain, a PDZ domain, a long proline-rich region that spans more than half the protein, and a sterile α motif (SAM) domain. This family includes SHANK1, SHANK2 and SHANK3, all of which localize to postsynaptic sites of excitatory synapses in the brain. SHANK2 protein lacks the ankyrin repeat.

Immunogen

SH3 and multiple ankyrin repeat domains protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SH3 and multiple ankyrin repeat domains 2 (SHANK2) protein is a multimodular scaffold protein present in the postsynaptic density (PSD). The PDZ domain of SHANK2 interacts with the C terminus of Phospholipase-β3 and brings it closer to Homer 1b for the regulation of mGluR-mediated calcium signal in the PSD region of neuronal synapses. Mutations in this gene have been linked to autism spectrum disorder and mental retardation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71536.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatemeh Hassani Nia et al.
Molecular psychiatry (2022-12-01)
Postsynaptic scaffold proteins such as Shank, PSD-95, Homer and SAPAP/GKAP family members establish the postsynaptic density of glutamatergic synapses through a dense network of molecular interactions. Mutations in SHANK genes are associated with neurodevelopmental disorders including autism and intellectual disability.
M Sheng et al.
Journal of cell science, 113 ( Pt 11), 1851-1856 (2000-05-12)
Shank proteins make up a new family of scaffold proteins recently identified through their interaction with a variety of membrane and cytoplasmic proteins. Shank polypeptides contain multiple sites for protein-protein interaction, including ankyrin repeats, an SH3 domain, a PDZ domain
Jong-Ik Hwang et al.
The Journal of biological chemistry, 280(13), 12467-12473 (2005-01-06)
Phospholipase C-beta isozymes that are activated by G protein-coupled receptors (GPCR) and heterotrimeric G proteins carry a PSD-95/Dlg/ZO-1 (PDZ) domain binding motif at their C terminus. Through interactions with PDZ domains, this motif may endow the PLC-beta isozyme with specific
Simone Berkel et al.
Nature genetics, 42(6), 489-491 (2010-05-18)
Using microarrays, we identified de novo copy number variations in the SHANK2 synaptic scaffolding gene in two unrelated individuals with autism-spectrum disorder (ASD) and mental retardation. DNA sequencing of SHANK2 in 396 individuals with ASD, 184 individuals with mental retardation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service