Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA008121

Sigma-Aldrich

Anti-GGT5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-γ-Glutamyltransferase 5 precursor antibody produced in rabbit, Anti-γ-Glutamyltransferase-like activity 1 antibody produced in rabbit, Anti-γ-Glutamyltranspeptidase 5 antibody produced in rabbit, Anti-GGT-rel antibody produced in rabbit, Anti-GGTLA1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GTSHVSVLGEDGSAVAATSTINTPFGAMVYSPRTGIILNNELLDLCERCPRGSGTTPSPVSGDRVGGAPGRCWPPVPGERSPSSMVPSILINKAQGSKLVIGGAGGELIISAVAQAIMSKLWLGFDLRAAIAAPILHVNSKG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GGT5(2687)

Immunogen

γ-Glutamyltransferase 5 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

GGT5 (γ-glutamyltransferase 5) gene encodes a protein belonging to the γ-glutamyl transpeptidase gene family. It cleaves the γ-glutamyl moiety of glutathione conjugates, such as antioxidants, inflammatory molecules, drug metabolites, and neuroactive compounds. It catalyzes the conversion of leukotriene C4 (LTC4) to leukotriene D4 (LTD4). It may be useful in the development of treatments for inflammatory diseases such as asthma and cardiovascular disease in humans. Increased levels of GGT have been observed in certain young Roma population in eastern Slovakia, and has been associated with metabolic syndrome. It functions as a pro-oxidant by generating reactive metabolite (CysGly) in the presence of iron and may be a part of a potential physiopathological oxidation system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73547

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Jarcuska et al.
Central European journal of public health, 22 Suppl, S43-S50 (2014-05-23)
Elevated gamma-glutamyl transpeptidase (GGT) is present approximately in half of all patients with non-alcoholic fatty liver disease (NAFLD). NAFLD is the liver manifestation of metabolic syndrome (MS). This study aimed to explore the relationship between GGT and MS or proinflammatory
Kevin Wei et al.
Nature, 582(7811), 259-264 (2020-06-06)
The synovium is a mesenchymal tissue composed mainly of fibroblasts, with a lining and sublining that surround the joints. In rheumatoid arthritis the synovial tissue undergoes marked hyperplasia, becomes inflamed and invasive, and destroys the joint1,2. It has recently been
M Enoiu et al.
Free radical biology & medicine, 29(9), 825-833 (2000-11-07)
It has been previously reported that the metabolism of reduced glutathione (GSH) by gamma-glutamyltranspeptidase (GGT) in the presence of chelated metals leads to free radical generation and lipid peroxidation (LPO). The present study demonstrates for the first time that an
Stephanie Wickham et al.
Analytical biochemistry, 414(2), 208-214 (2011-03-31)
Gamma-glutamyl compounds include antioxidants, inflammatory molecules, drug metabolites, and neuroactive compounds. Two cell surface enzymes that metabolize gamma-glutamyl compounds have been identified: gamma-glutamyl transpeptidase (GGT1) and gamma-glutamyl leukotrienase (GGT5). There is controversy in the literature regarding the substrate specificity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service