Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA007979

Sigma-Aldrich

Anti-FKBP15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FKBP133, Anti-KIAA0674, Anti-PPP1R76, Anti-WAFL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KHSAGNSMLIPSMSVTMETSMIMSNIQRIIQENERLKQEILEKSNRIEEQNDKISELIERNQRYVEQSNLMMEKRNNSLQTATENTQARVLHAEQEKAKVTEELAAATAQVSHLQLKMTAHQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FKBP15(23307)

Immunogen

FK506-binding protein 15 recombinant protein epitope signature tag (PrEST)

Application

Anti-FKBP15 antibody produced in rabbit has been used for co-immunoprecipitation.
Anti-FKBP15 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

FKBP15 (FK506 binding protein 15) gene encodes a 133kDa protein that forms a tripartite complex with the retromer cargo-selective complex (CSC) and the WASH (Wiskott–Aldrich syndrome homologue) complex, a multimeric protein complex that is involved in the regulation of tubule dynamics at endosomes. It facilitates or stabilizes this tripartite complex. FKBP15 may function in the regulation of actin dynamics at the endosome. This protein has been implicated in inflammatory bowel disease (Ulcerative colitis).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71229

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthew Nj Seaman et al.
Communicative & integrative biology, 7, e29483-e29483 (2014-07-30)
The retromer complex mediates endosomal protein sorting by concentrating membrane proteins (cargo) into nascent tubules formed through the action of sorting nexin (SNX) proteins. The WASH complex is recruited to endosomes by binding to the VPS35 subunit of retromer and
Michael E Harbour et al.
The Biochemical journal, 442(1), 209-220 (2011-11-11)
The retromer complex is a conserved endosomal protein sorting complex that sorts membrane proteins into nascent endosomal tubules. The recognition of membrane proteins is mediated by the cargo-selective retromer complex, a stable trimer of the Vps35 (vacuolar protein sorting 35)
You Fu Pan et al.
International journal of biological sciences, 6(2), 163-171 (2010-04-09)
Ulcerative colitis (UC) is one of the major forms of inflammatory bowel disease with unknown cause. A molecular marker, WAFL, has recently been found to be up-regulated in the inflamed colonic mucosa of UC patients. Towards understanding biological function of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service