Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA007923

Sigma-Aldrich

Anti-MAGI3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-membrane-associated guanylate kinase-related 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

LKVGDHISAVNGQSIVELSHDNIVQLIKDAGVTVTLTVIAEEEHHGPPSGTNSARQSPALQHRPMGQSQANHIPGDRSALEGEIGKDVSTSYRHSWSDHKHLAQPDTAVISVVGSRHNQNLGCYPVELERGPRGFGFSLRG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAGI3(260425)

Immunogen

Membrane-associated guanylate kinase, WW and PDZ domain-containing protein 3 (Membrane-associated guanylate kinase inverted 3) (MAGI-3)

Application

Anti-MAGI3 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry[1] against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

MAGI3 (membrane associated guanylate kinase, WW and PDZ domain containing 3) gene encodes a membrane-associated guanylate kinase that contains multi-PDZ domains. It is a scaffolding protein that is found at cell-cell junctions. It interacts with frizzled-4 and -7 (seven-transmembrane proteins involved in PCP/ planar cell polarity pathway) and Ltap (a mouse homolog of the Drosophila PCP protein, stbm) to form a complex. This complex is found to activate JNK signaling cascade. It binds to the (PDZ) recognition motif (TVV) that is present at the C-terminal of the transforming growth factor-α precursor (proTGFα) and facilitates efficient trafficking of TGFα to the cell surface in polarized epithelial cells. MAGI3 binds to the PDZ domain-binding site of PTEN/MMAC, a tumor suppressor, and modulates the kinase activity of AKT/PKB (protein kinase B). It binds to the PDZ-binding domain of LPA2 (lysophosphatidic acid 2) and regulates the ability of LPA2 to activate Erk (extracellular signal regulated kinase) and RhoA.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71112

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huanchun Zhang et al.
Cellular signalling, 19(2), 261-268 (2006-08-15)
Lysophosphatidic acids (LPA) exert multiple biological effects through specific G protein-coupled receptors. The LPA-activated receptor subtype LPA(2) contains a carboxyl-terminal motif that allows interaction with PDZ domain-containing proteins, such as NHERF2 and PDZ-RhoGEF. To identify additional interacting partners of LPA(2)
Jeffrey L Franklin et al.
Experimental cell research, 303(2), 457-470 (2005-01-18)
The cytoplasmic domain of the transforming growth factor-alpha precursor (proTGFalpha) contains a C-terminal PSD-95/SAP90, Discs Large, and Zona Occludens-1 (PDZ) recognition motif (TVV). By yeast two-hybrid screening of a mouse embryo library, we have found that a third member of
Ryoji Yao et al.
Oncogene, 23(36), 6023-6030 (2004-06-15)
A seven-transmembrane protein, frizzled, has been implicated in a planar cell polarity (PCP) pathway as well as the canonical Wnt signaling pathway. Although both pathways require a cytoplasmic protein, dishevelled, the molecular mechanism by which frizzled regulates intracellular signaling remains
Y Wu et al.
The Journal of biological chemistry, 275(28), 21477-21485 (2000-04-05)
PTEN/MMAC is a phosphatase that is mutated in multiple human tumors. PTEN/MMAC dephosphorylates 3-phosphorylated phosphatidylinositol phosphates that activate AKT/protein kinase B (PKB) kinase activity. AKT/PKB is implicated in the inhibition of apoptosis, and cell lines and tumors with mutated PTEN/MMAC
Gabriel Belem de Andrade et al.
Molecular vision, 20, 1422-1433 (2014-10-30)
The Down syndrome cell adhesion molecule (Dscam) gene is required for normal dendrite arborization and lamination in the mouse retina. In this study, we characterized the developmental localization of the DSCAM protein to better understand the postnatal stages of retinal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service