Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA007637

Sigma-Aldrich

Anti-NTRK2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BDNF/NT-3 growth factors receptor precursor antibody produced in rabbit, Anti-GP145-TrkB antibody produced in rabbit, Anti-Neurotrophic tyrosine kinase receptor type 2 antibody produced in rabbit, Anti-Trk-B antibody produced in rabbit, Anti-TrkB tyrosine kinase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

FAPTITFLESPTSDHHWCIPFTVKGNPKPALQWFYNGAILNESKYICTKIHVTNHTEYHGCLQLDNPTHMNNGDYTLIAKNEYGKDEKQISAHFMGWPGIDDGANPNYPDVIYEDYGTAANDIGDTTNRSNEIP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NTRK2(4915)

General description

NTRK2 (neurotrophic tyrosine kinase, receptor, type 2) gene is mapped to human chromosome 9q22.1 and spans a length of 353kb. It consists of 21 exons that are arranged in six different blocks. This protein is composed of the promoter region, the extracellular domain and the juxtamembrane region, intracellular juxtamembane domain, followed by the tyrosine kinsase domain. The C-terminus of the tyrosine kinase domain and a γ phospholipase C binding site are encoded by exons 20 and 21.

Immunogen

BDNF/NT-3 growth factors receptor precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NTRK2 (neurotrophic tyrosine kinase, receptor, type 2) gene encodes a membrane-bound receptor belonging to the neurotrophic tyrosine receptor kinase (NTRK) family. It undergoes autophosphorylation on binding to neurotrophin and phosphorylates the members of MAPK pathway. NTRK2 is expressed in hypothalamic neurons that are involved in the control of food intake and energy expenditure. Polymorphism in this gene is linked to eating disorders leading to obesity. It regulates short- and long-term synaptic function and neural plasticity along with its ligand brain-derived neurotropic factor. Alterations in this gene can cause major depressive disorder (MDD).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71430

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Fleury et al.
Frontiers in immunology, 12, 606861-606861 (2021-03-02)
Platelets and neurons share many similarities including comparable secretory granule types with homologous calcium-dependent secretory mechanisms as well as internalization, sequestration and secretion of many neurotransmitters. Thus, platelets present a high potential to be used as peripheral biomarkers to reflect
C Dong et al.
Molecular psychiatry, 14(12), 1105-1118 (2009-10-22)
We studied seven genes that reflect events relevant to antidepressant action at four sequential levels: (1) entry into the brain, (2) binding to monoaminergic transporters, and (3) distal effects at the transcription level, resulting in (4) changes in neurotrophin and
M Ribases et al.
Molecular psychiatry, 10(9), 851-860 (2005-04-20)
Anorexia nervosa (AN) and bulimia nervosa (BN) are eating disorders (ED) with complex genetic and environmental components. Genetic studies and animal models support the participation of brain-derived neurotrophic factor (BDNF) in the vulnerability to AN and BN. We investigated the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service