Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA007458

Sigma-Aldrich

Anti-PNCK antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CaM kinase I β antibody produced in rabbit, Anti-CaM kinase IB antibody produced in rabbit, Anti-CaM-KI β antibody produced in rabbit, Anti-CaMKI-β antibody produced in rabbit, Anti-Calcium/calmodulin-dependent protein kinase type 1B antibody produced in rabbit, Anti-Pregnancy up-regulated non-ubiquitously expressed CaM kinase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

MLLLKKHTEDISSVYEIRERLGSGAFSEVVLAQERGSAHLVALKCIPKKALRGKEALVENEIAVLRRISHPN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PNCK(139728)

General description

PNCK (pregnancy up-regulated nonubiquitous CaM kinase) gene encodes a serine/threonine kinase that shares homology with members of the CaM kinase family. The molar mass of this protein is 38kDa. The gene is mapped to human chromosome Xq28 that is linked to multiple distinct mental retardation syndromes. The expression levels of this protein increases during intermediate and late stages of murine fetal development and is specifically expressed in certain tissues in brain, uterus, ovary, and testis. Its expression is highest in murine late pregnancy during mammary development.

Immunogen

Calcium/calmodulin-dependent protein kinase type 1B recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PNCK (pregnancy up-regulated nonubiquitous CaM kinase) is a Ca2+/calmodulin-dependent kinase that induces degradation of ligand-independent epidermal growth factor receptor. It may play a role in the development of the central nervous system. It may also function in mammary development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71420

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanji Xu et al.
Journal of Cancer, 10(27), 6925-6932 (2019-12-17)
Purpose: Recent studies indicate that pregnancy upregulated non-ubiquitous calmodulin kinase (PNCK) is significantly up-regulated in breast and renal carcinomas. However, the expression profile and its biological relevance of PNCK in nasopharyngeal carcinoma (NPC) have not been elucidated. Methods: The expression
Yoon Ah Cho et al.
Cancer genomics & proteomics, 17(6), 747-755 (2020-10-26)
Pregnancy up-regulated non-ubiquitous calmodulin kinase (PNCK) is a member of calmodulin kinase, and overexpression of PNCK with involvement in carcinogenesis have been reported in HER-2 amplified breast cancer, clear cell renal cell carcinoma and nasopharygeal carcinoma. However, the expression of
H P Gardner et al.
Cancer research, 60(19), 5571-5577 (2000-10-18)
While screening for protein kinases expressed in the murine mammary gland, we identified previously a Ca2+/calmodulin-dependent kinase, Pnck, that is most closely related to CaMKI. In this report, we show that Pnck is temporally regulated during murine mammary development with
Tushar B Deb et al.
American journal of physiology. Cell physiology, 300(5), C1139-C1154 (2011-02-18)
We have recently described a novel role for pregnancy-upregulated non-ubiquitous calmodulin kinase (Pnck) in the induction of ligand-independent epidermal growth factor receptor (EGFR) degradation (Deb TB, Coticchia CM, Barndt R, Zuo H, Dickson RB, and Johnson MD. Am J Physiol
H P Gardner et al.
Genomics, 63(2), 279-288 (2000-02-16)
Calcium is an important second messenger in eukaryotic cells. Many of the effects of calcium are mediated via its interaction with calmodulin and the subsequent activation of Ca(2+)/calmodulin-dependent (CaM) kinases. CaM kinases are involved in a wide variety of cellular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service