Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA007455

Sigma-Aldrich

Anti-MRPL2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-39S ribosomal protein L2, mitochondrial precursor antibody produced in rabbit, Anti-L2mt antibody produced in rabbit, Anti-MRP-L2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PAPSLFPAAQMMNNGLLQQPSALMLLPCRPVLTSVALNANFVSWKSRTKYTITPVKMRKSGGRDHTGRIRVHGIGGGHKQRYRMIDFLRFRPEETKSGPFEEKVIQVRYDP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPL2(51069)

Immunogen

39S ribosomal protein L2, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MRPL2 (mitochondrial ribosomal protein L2) gene encodes a protein belonging to the EcoL2 ribosomal protein family. It is a component of the larger 39S subunit of the human mitochondrial ribosome. The ribosomal proteins are encoded by nuclear genes and they function in protein synthesis within the mitochondrion. The gene is mapped to human chromosome 6p21.1 and is a candidate gene for spinocerebellar ataxia with blindness and deafness.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71805

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vikas Yadav et al.
Yeast (Chichester, England), 28(1), 81-91 (2010-10-23)
Chlorophenols are a class of chemicals commonly used in preservatives, disinfectants, algaecides, herbicides and pesticides. However, there is a growing evidence that these compounds are a threat to human health. This is alarming as many chlorophenols are common pollutants found
E C Koc et al.
The Journal of biological chemistry, 276(47), 43958-43969 (2001-09-12)
Identification of all the protein components of the large subunit (39 S) of the mammalian mitochondrial ribosome has been achieved by carrying out proteolytic digestions of whole 39 S subunits followed by analysis of the resultant peptides by liquid chromatography
Thomas W O'Brien et al.
Gene, 354, 147-151 (2005-05-24)
The ancestral mitochondrial ribosome (70S) underwent major structural remodeling during the evolution of mammalian mitochondrial ribosomes (55S). Despite the loss of nearly half their RNA, 55S ribosomes are actually larger than bacterial ribosomes because of all the extra proteins they

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service