Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA005647

Sigma-Aldrich

Anti-DIAPH2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DRF2 antibody produced in rabbit, Anti-Diaphanous-related formin-2 antibody produced in rabbit, Anti-Protein diaphanous homolog 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Check Cart for Availability


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

GGSEEPGGGRSNKRSAGNRAANEEETKNKPKLNIQIKTLADDVRDRITSFRKSTVKKEKPLIQHPIDSQVAMSEFPAAQPLYDERSLNLSEKEVLDLFEKMMEDMNLNEEKKAPLRNKDFTTKREMVVQYISAT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DIAPH2(1730)

General description

Diaphanous-related formin 2 (DIAPH2) is part of the diaphanous subfamily of the formin homology family of proteins. It is a nuclear protein. A defect in this gene leads to premature ovarian failure. The gene DIAPH2 is mapped to human chromosome Xq21.33.

Immunogen

Protein diaphanous homolog 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Diaphanous-related formin 2 (DIAPH2) interacts specifically with the distal regulatory element (DRE) of the human skeletal α-actin gene between positions -1260 and -1193, the binding site being an A/T-rich sequence element. The DRF-2 also binds well to the muscle creatine kinase (MCK) muscle-specific regulatory factor, MEF-2 binding site and to the A/T-rich regulatory element of the skeletal muscle fast-twitch troponin C gene. This troponin C site competes for DRF-2 driven expression of the skeletal α-actin gene in C2 cells. The DRF-2 site alone does not activate transcription in muscle cells when linked to the SV40 promoter. DRF-2 binding element is a MEF-2 binding site that is required but insufficient for regulation of muscle-specific skeletal α-actin gene expression by the DRE. Therefore, muscle-specific regulation of the human skeletal α-actin gene requires interactions between the other elements of the composite DRE enhancer with the protein-DNA complex formed by DRF-2.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74314

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bixi Zhang et al.
Frontiers in molecular biosciences, 9, 910950-910950 (2023-01-03)
The diaphanous-related formin subfamily includes diaphanous homolog 1 (DIAPH1), DIAPH2, and DIAPH3. DIAPHs play a role in the regulation of actin nucleation and polymerization and in microtubule stability. DIAPH3 also regulates the assembly and bipolarity of mitotic spindles. Accumulating evidence
Rita Genesio et al.
Molecular cytogenetics, 8, 50-50 (2015-07-16)
Primary ovarian insufficiency (POI) is defined as a primary ovarian defect characterized by absent menarche (primary amenorrhea), a decrease in the initial primordial follicle number, high follicle-stimulating hormone (FSH) levels and hypoestrogenism. Although the etiology of a majority of POI
G E Muscat et al.
Gene expression, 2(2), 111-126 (1992-01-01)
The tissue-specific distal promoter of the human skeletal alpha-actin gene (-1282 to -708) induces transcription in myogenic cells approximately 10-fold and, with the most proximal promoter domain (-153 to -87), it synergistically increases transcription 100-fold (Muscat and Kedes 1987). We
Saskia S Grueb et al.
Scientific reports, 9(1), 5352-5352 (2019-03-31)
In this study, we analyzed the functional role of the formin Drosophila Homologue of Diaphanous2 (Diaph2) in colorectal cancer cells. We show that stable down-regulation of Diaph2 expression in HT29 cells decreased chromosome alignment and the velocity of chromosome movement
Kristine Oevel et al.
eLife, 12 (2024-03-19)
Neurotransmission at synapses is mediated by the fusion and subsequent endocytosis of synaptic vesicle membranes. Actin has been suggested to be required for presynaptic endocytosis but the mechanisms that control actin polymerization and its mode of action within presynaptic nerve

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service