Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA005465

Sigma-Aldrich

Anti-FMN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti- LD, Anti-DKFZP686C2281, Anti-FLJ45135, Anti-FMN, Anti-MGC125288, Anti-MGC125289

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

GFGSFETAFKDTGLGREVLPPDCSSTEAGGDGIRRPPSGLEHQQTGLSESHQDPEKHPEAEKDEMEKPAKRTCKQKPVSKVVAKVQDLSSQVQRVVKTHSKGKETIAIRPAAHAEFVPKADLLT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FMN1(342184)

General description

Formin-1 (FMN1) belongs to the family of non-FDD type formin homology proteins. This gene is localized to chromosome 15q13.3, and produces two FMN1 isoforms- FMN1isoform 1 and FMN1 isoform 2. This is a result of alternate splicing of the alternative promoter type. This protein consists of FH1, FH2, FMH1 and FMH2 domains.

Immunogen

FMN1_HUMAN Isoform 2 of Q68DA7 - Homo sapiens recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Formin-1 (FMN1) is responsible for the polymerization of actin filaments. It interacts with α-catenin and moves to nascent adherens junctions, where it nucleates actin filaments. Thus, it plays an essential role in the formation of cell-cell junctions by mediating adherens junction formation. Mutations in this gene in mouse, give rise to recessively inherited murine limb deformity, which is characterized by limb deformity or renal aplasia. Studies have shown that mutation in FMN1 gene in humans gives rise to oligosyndactyly, renal defects, radioulnar synostosis and hearing loss. This phenotype is similar to mice limb deformity. Duplication in this gene has been found in a patient with Cenani-Lenz-like oligosyndactyly of the hands.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70053

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascale Monzo et al.
Developmental cell, 56(20), 2841-2855 (2021-09-25)
Glioblastoma are heterogeneous tumors composed of highly invasive and highly proliferative clones. Heterogeneity in invasiveness could emerge from discrete biophysical properties linked to specific molecular expression. We identified clones of patient-derived glioma propagating cells that were either highly proliferative or
R L Maas et al.
American journal of human genetics, 48(4), 687-695 (1991-04-01)
The murine limb deformity (ld) locus resides on mouse chromosome 2 and gives rise to a recessively inherited, characteristic limb deformity/renal aplasia phenotype. In this locus in the mouse, a gene, termed the "formin" gene, has been identified which encodes
Agnieszka Kobielak et al.
Nature cell biology, 6(1), 21-30 (2003-12-03)
During epithelial sheet formation, linear actin cables assemble at nascent adherens junctions. This process requires alpha-catenin and actin polymerization, although the underlying mechanism is poorly understood. Here, we show that formin-1 interacts with alpha-catenin, localizes to adherens junctions and nucleates
Boyan Ivanov Dimitrov et al.
Journal of medical genetics, 47(8), 569-574 (2010-07-09)
Limb development is a complex process requiring proper spatio-temporal expression of a network of limb specific morphogens. Grem1 and Fmn1 play an important role in mouse and chick limb development. The mouse limb deformity (ld) phenotype with digit reduction, syndactyly
Masuko Katoh et al.
International journal of molecular medicine, 13(3), 473-478 (2004-02-10)
Formin homology proteins with FH1 and FH2 domains are signaling effectors for assembly and polarization of actin filaments. FH1 is the binding domain for Profilin, SRC, EMS1/Cortactin, FNBP1, FNBP2, FNBP3, FNBP4 and WBP4/Fbp21, while FH2 is the actin-filament modification domain.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service