Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA005448

Sigma-Aldrich

Anti-RAD17 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cell cycle checkpoint protein RAD17 antibody produced in rabbit, Anti-RF-C/activator 1 homolog antibody produced in rabbit, Anti-hRad17 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$555.00

$555.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$555.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$555.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

SKEHGIQVQEWINPVLPDFQKDDFKGMFNTESSFHMFPYQSQIAVFKEFLLRATKYNKLQMLGDDLRTDKKIILVEDLPNQFYRDSHTLHEVLRKYVRIGRCPLIFIISDSLSGDNNQRLLFPKEIQEECSISNISFNPVAP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD17(5884)

General description

Cell cycle checkpoint protein RAD17 is encoded by the gene hRAD17, in humans. hRAD17 is a 69kDa protein, which is made of 590 amino acids. RAD17 was first identified in Schizosaccharomyces pombe, and its homolog in humans is hRAD17. Human RAD17 gene is localized to the chromosomal region 5q13.

Immunogen

Cell cycle checkpoint protein RAD17 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

RAD17 is a DNA damage cell cycle checkpoint protein. RAD17 forms a complex with four small replication factor c (RFC) subunits namely, Rfc2, Rfc3, Rfc4 and Rfc5. This complex, called RFC-related complex, acts as a clamp loader and detects DNA damage. Once DNA damage is detected, RFC-related complex recruits 9-1-1 complex (Rad9, Rad1 and Hus1). This, in turn, activates ATM, ATR, Chk1 and Chk2 kinases. Therefore, RAD17 plays an essential role in maintaining genomic stability. RAD17 might be a potential tumor suppressor gene and may have a role in the progression of head and neck squamous cell carcinoma (HNSCC) by contributing to genomic instability.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86856

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Li et al.
Oncogene, 18(9), 1689-1699 (1999-04-20)
The RAD17 gene product of S. Pombe is an essential component of the checkpoint control pathway which responds to both DNA damage and disruption of replication. We have identified a human cDNA that encodes a polypeptide which is structurally conserved
Xin Wang et al.
Genes & development, 17(8), 965-970 (2003-04-04)
Cell cycle checkpoints are critical for genomic stability. Rad17, a component of the checkpoint clamp loader complex (Rad17/Rfc2-5), is required for the response to DNA damage and replication stress. To explore the role of Rad17 in the maintenance of genomic
Hiroyuki Niida et al.
Mutagenesis, 21(1), 3-9 (2005-11-30)
DNA damage is a common event and probably leads to mutation or deletion within chromosomal DNA, which may cause cancer or premature aging. DNA damage induces several cellular responses including DNA repair, checkpoint activity and the triggering of apoptotic pathways.
Ming Zhao et al.
Head & neck, 30(1), 35-42 (2007-07-28)
DNA repair genes play a critical role in maintaining genome stability and have been implicated in tumorigenesis. Head and neck squamous cell carcinoma (HNSCC) often shows chromosomal instability. We examined the expression of human RAD17, a DNA damage cell cycle

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service