Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA005445

Sigma-Aldrich

Anti-TULP4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Tubby superfamily protein antibody produced in rabbit, Anti-Tubby-like protein 4 antibody produced in rabbit, Anti-Tubby-related protein 4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Check Cart for Availability


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$598.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

VLLHESDGVLGMSWNYPIFLVEDSSESDTDSDDYAPPQDGPAAYPIPVQNIKPLLTVSFTSGDISLMNNYDDLSPTVIRSGLKEVVAQWCTQGDLLAVAGMERQTQLGELPNGPLLKSAMVKFYNVRGEHIFTLDTLVQRPIISICWGH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TULP4(56995)

General description

Tubby like protein 4 (TULP4) belongs to the tubby superfamily protein (TUSP) found in both humans and mouse. TULP4 is also called TUSP and in humans the gene spans a length of 11,127bp. TULP4 protein is composed of 1544 amino acids. Human TULP4 gene is localized to chromosome 6q25-q26. TULP4 protein contains a highly conserved tubby domain at its C-terminal. It is expressed in testis, brain, skeletal muscle, and kidney.

Immunogen

Tubby-related protein 4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In mammals, tubby like proteins (TULP) regulate the function of neuronal cells during development and postdifferentiation. However, the exact role of TULP4 is yet not known.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70743

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Q Z Li et al.
Gene, 273(2), 275-284 (2001-10-12)
We report here the cloning and characterization of a novel gene belonging to the tubby superfamily proteins (TUSP) in mouse and human. The mouse Tusp cDNA is 9120 bp in length and encodes a deduced protein of 1547 amino acids
Chia-Ping Lai et al.
Plant physiology, 134(4), 1586-1597 (2004-04-06)
In mammals, TUBBY-like proteins play an important role in maintenance and function of neuronal cells during postdifferentiation and development. We have identified a TUBBY-like protein gene family with 11 members in Arabidopsis, named AtTLP1-11. Although seven of the AtTLP genes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service