Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA004171

Sigma-Aldrich

Anti-AUH antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AU-binding protein/enoyl-CoA hydratase, Anti-AU-specific RNA-binding enoyl-CoA hydratase, Anti-Methylglutaconyl-CoA hydratase, mitochondrial precursor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$598.00

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μL
$598.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$598.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

SAWLCPGLRLPGSLAGRRAGPAIWAQGWVPAAGGPAPKRGYSSEMKTEDELRVRHLEEENRGIVVLGINRAYGKNSLSKNLIKMLSKAVDALKSDKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AUH(549)

General description

AU RNA binding protein/enoyl-CoA hydratase (AUH) is a bifunctional protein that encodes an RNA binding protein with intrinsic enzymatic activity.

Immunogen

Methylglutaconyl-CoA hydratase, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

AUH is mainly associated with two major activities: RNA binding activity and catalysis of the hydration reaction of 2-trans-enoyl-coenzyme A (enoyl-CoA). The AUH trimer molecule forms dimer by binding to AUUU motif of a long RNA strand. It functions as a 3-hydroxy-3-methylglutaconyl-CoA dehydratase in the leucine degradation pathway. Mutation in AUH affects the leucine degradation pathway. In deficiency of 3-methylglutaconyl-CoA hydratase, one of the key enzymes of leucine catametabolism cascade which is encoded by AUH causes 3-Methylglutaconic aciduria type I, an autosomal recessive disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86719

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lodewijk IJlst et al.
American journal of human genetics, 71(6), 1463-1466 (2002-11-16)
3-Methylglutaconic aciduria type I is an autosomal recessive disorder clinically characterized by various symptoms ranging from delayed speech development to severe neurological handicap. This disorder is caused by a deficiency of 3-methylglutaconyl-CoA hydratase, one of the key enzymes of leucine
Kazuki Kurimoto et al.
Proteins, 75(2), 360-372 (2008-10-03)
The human AU RNA binding protein/enoyl-Coenzyme A hydratase (AUH) is a 3-hydroxy-3-methylglutaconyl-CoA dehydratase in the leucine degradation pathway. It also possesses an RNA-binding activity to AUUU repeats, which involves no known conserved RNA-binding domains and is seemingly unrelated to the
K Kurimoto et al.
Structure (London, England : 1993), 9(12), 1253-1263 (2001-12-12)
The AU binding homolog of enoyl-CoA hydratase (AUH) is a bifunctional protein that has two distinct activities: AUH binds to RNA and weakly catalyzes the hydration of 2-trans-enoyl-coenzyme A (enoyl-CoA). AUH has no sequence similarity with other known RNA binding
J Nakagawa et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(6), 2051-2055 (1995-03-14)
AU-rich elements within the 3' untranslated region of transcripts of lymphokines and some protooncogenes serve as signal for rapid mRNA degradation. By using an AUUUA matrix, we have affinity-purified a 32-kDa protein, microsequenced it, and cloned the corresponding cDNA. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service